Search
Search
| Gene Symbol: Casq1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159594.1 | NP_001153066.1 | 5 - 6 | 814 | 75 | |
| GenBank mRNA | FM056035.1 | - | 2 - 3 | 119 | 75 | |
| FQ216689.1 | - | 5 - 6 | 391 | 75 | ||
| FQ224875.1 | - | 6 - 7 | 641 | 75 | ||
Set Membership: |
|