Search
Search
| Gene Symbol: Smad3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013095.3 | NP_037227.1 | 2 - 3 | 601 | 103 | |
| GenBank mRNA | BC064437.1 | - | 601 | 5599910 | 103 | |
| U66479.1 | - | 2 - 3 | 410 | 103 | ||
Set Membership: |
|