Search
Search
| Gene Symbol: Izumo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017514.1 | NP_001017514.1 | 7 - 8 | 898 | 78 | |
| XM_008759406.1 | XP_008757628.1 | 6 - 7 | 1040 | 78 | ||
| GenBank mRNA | AB195683.1 | - | 6 - 7 | 662 | 78 | |
| BC079244.1 | - | 7 - 8 | 898 | 7132791 | 78 | |
Set Membership: |
|