Search
Search
| Gene Symbol: Renbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031095.1 | NP_112357.1 | 2 - 3 | 198 | 88 | |
| XM_006229615.2 | XP_006229677.1 | 3 - 4 | 320 | 88 | ||
| GenBank mRNA | D10233.1 | - | 2 - 3 | 198 | 88 | |
Set Membership: |
|