Search
Search
| Gene Symbol: Acer2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107943.1 | NP_001101413.1 | 1 - 2 | 202 | 63 | |
| XM_006238363.2 | XP_006238425.1 | 1 - 2 | 244 | 63 | ||
| GenBank mRNA | FQ229191.1 | - | 1 - 2 | 177 | 63 | |
Set Membership: |
|