Search
Search
| Gene Symbol: TEX30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013991669.2 | XP_013847123.1 | 216 | 136 | ||
| XM_021065893.1 | XP_020921552.1 | 213 | 136 | |||
| GenBank mRNA | AY609434.1 | - | 2 - 3 | 103 | 136 | |
Set Membership: |
|