Search
Search
| Gene Symbol: RNF19A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038006.1 | NP_001033095.1 | 1 - 2 | 673 | 82 | |
| XM_005662947.3 | XP_005663004.1 | 1298 | 82 | |||
| XM_013996572.2 | XP_013852026.1 | 1069 | 82 | |||
| XM_013996573.2 | XP_013852027.1 | 1157 | 82 | |||
| XM_013996574.2 | XP_013852028.1 | 1168 | 82 | |||
| GenBank mRNA | DQ091176.1 | - | 1 - 2 | 673 | 82 | |
Set Membership: |
|