Search
Search
| Gene Symbol: SGO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005672058.3 | XP_005672115.1 | 4353 | 113 | ||
| XM_005672059.3 | XP_005672116.1 | 4221 | 113 | |||
| XM_013984371.2 | XP_013839825.1 | 4030 | 113 | |||
| XM_021074895.1 | XP_020930554.1 | 4174 | 113 | |||
| XM_021074897.1 | XP_020930556.1 | 4122 | 113 | |||
| GenBank mRNA | AK394404.1 | - | 2 - 3 | 695 | 113 | |
Set Membership: |
|