Search
Search
| Gene Symbol: GK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003483257.4 | XP_003483305.2 | 1203 | 108 | ||
| XM_021069567.1 | XP_020925226.1 | 1411 | 108 | |||
| XM_021069569.1 | XP_020925228.1 | 1055 | 108 | |||
| XM_021069570.1 | XP_020925229.1 | 1221 | 108 | |||
| GenBank mRNA | AK395853.1 | - | 11 - 12 | 1121 | 108 | |
Set Membership: |
|