Search
Search
| Gene Symbol: PHF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021088179.1 | XP_020943838.1 | 252 | 58 | ||
| XM_021088180.1 | XP_020943839.1 | 253 | 58 | |||
| XM_021088181.1 | XP_020943840.1 | 253 | 58 | |||
Set Membership: |
|