Search
Search

cpm| Gene Symbol: cpm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011270.1 | NP_001011270.1 | 5 - 6 | 640 | 80 | |
| XM_012958933.2 | XP_012814387.1 | 6 - 7 | 1046 | 80 | ||
| XM_012958934.2 | XP_012814388.1 | 5 - 6 | 853 | 80 | ||
| XM_012958935.2 | XP_012814389.1 | 5 - 6 | 1580 | 80 | ||
| XM_012958937.2 | XP_012814391.1 | 4 - 5 | 497 | 80 | ||
| XM_018092056.1 | XP_017947545.1 | 6 - 7 | 1042 | 80 | ||
| GenBank mRNA | BC087979.1 | - | 5 - 6 | 640 | 80 | |
Set Membership: |
|