Search
Search

kat14| Gene Symbol: kat14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079210.2 | NP_001072678.1 | 9 - 10 | 2384 | 93 | |
| GenBank mRNA | BC124007.1 | - | 8 - 9 | 2015 | 93 | |
| BC136099.1 | - | 9 - 10 | 2384 | 93 | ||
Set Membership: |
|