Search
Search

phb4| Gene Symbol: phb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111499.1 | NP_001104969.1 | 369 | 84 | ||
| GenBank mRNA | AF236371.1 | - | 405 | 84 | ||
| AY109093.2 | - | 369 | 84 | |||
Set Membership: |
|