Search Thermo Fisher Scientific
In this assay, the modified thymidine analog EdU (5-ethynyl-2'-deoxyuridine, a nucleoside analog of thymidine) is efficiently incorporated into newly synthesized DNA and fluorescently labeled with a bright, photostable Alexa Fluor® dye in a fast, highly-specific, mild click reaction
| Reaction components* |
Number of coverslips
| ||||||
|---|---|---|---|---|---|---|---|
| 1 | 2 | 4 | 5 | 10 | 25 | 50 | |
| 1X Click-iT® EdU reaction buffer | 430 µL | 860 µL | 1.8 mL | 2.2 mL | 4.3 mL | 10.7 mL | 21.4 mL |
| CuSO4 (Component E) | 20 µL | 40 µL | 80 µL | 100 µL | 200 µL | 500 µL | 1 mL |
| Alexa Fluor® azide | 1.2 µL | 2.5 µL | 5 µL | 6 µL | 12.5 µL | 31 µL | 62 µL |
| 1X Click-iT® EdU buffer additive | 50 µL | 100 µL | 200 µL | 250 µL | 500 µL | 1.25 mL | 2.5 mL |
| Total volume | 500 µL | 1 mL | 2 mL | 2.5 mL | 5 mL | 12.5 mL | 25 mL |
| *Note: Add the ingredients in the order listed in the table. | |||||||
| Hoechst® 33342 | Alexa Fluor® 488 | Alexa Fluor® 555 | Alexa Fluor® 594 | Alexa Fluor® 647 | |
|---|---|---|---|---|---|
| Excitation/Emission (in nm) | 350/461 | 495/519 | 555/565 | 590/615 | 650/670 |
| Standard filter set | DAPI | FITC | RFP | TRITC | Cy®5 |