Click-iT™ EdU Cell Proliferation Kit for Imaging, Alexa Fluor™ 594 dye
Invitrogen™

Click-iT™ EdU Cell Proliferation Kit for Imaging, Alexa Fluor™ 594 dye

The Click-iT EdU Cell Proliferation Kit for Imaging is a superior alternative to traditional proliferation assays that is optimized forRead more
Have Questions?
Catalog NumberQuantity
C103391 kit
Catalog number C10339
Price (TWD)
27,300.00
Online offer
Ends: 31-Dec-2026
39,000.00
Save 11,700.00 (30%)
Each
Quantity:
1 kit
Price (TWD)
27,300.00
Online offer
Ends: 31-Dec-2026
39,000.00
Save 11,700.00 (30%)
Each
The Click-iT EdU Cell Proliferation Kit for Imaging is a superior alternative to traditional proliferation assays that is optimized for fluorescence microscopy applications. In this assay the modified thymidine analogue EdU is efficiently incorporated into newly synthesized DNA and fluorescently labeled with a bright, photostable Alexa Fluor™ dye in a fast, highly-specific click reaction. This fluorescent labeling of proliferating cells is accurate and compatible with antibody methods due to the mild click protocol.

• Simple—works the first time, every time, in less time
• Efficient—no denaturation steps or harsh treatment required
• Content-rich results—better preservation of cell morphology, antigen structure, and DNA integrity
• Consistent—not dependent on variable antibody lots for detection

The most accurate proliferation-detection methods are based on the incorporation and measurement of nucleoside analogues in newly synthesized DNA, with bromodeoxyuridine (BrdU) a commonly used analogue. BrdU-labeled DNA is quantitated using anti-BrdU antibodies following DNA denaturation by harsh methods (HCl, heat, or enzymes) to expose the BrdU molecules. This step is time consuming and difficult to perform consistently. The harsh treatment can also adversely effect sample integrity and quality, which makes co-staining with other antibodies challenging.

Superior Proliferation Methodology
The Click-iT EdU Cell Proliferation Kit for Imaging provides a superior alternative to BrdU assays for measuring cell proliferation. EdU (5-ethynyl-2'-deoxyuridine) is a nucleoside analog of thymidine and is incorporated into DNA during active DNA synthesis. With Click-iT™ EdU, mild fixation and detergent permeabilization is sufficient for the small molecule-based Click-iT™ EdU detection reagent to gain access to the DNA. As a consequence, the Click-iT™ EdU imaging kit is not only easy to use, but more accurate and compatible with cell cycle analysis and other intracellular or extracellular targets for truly content-rich results.

This kit is optimized for fluorescence microscopy applications; visit the Click-iT™ technology area of our website for kits designed for high-content imaging, fluorescence microplate, or flow cytometry platforms.

Learn more about Click-iT technology >
Find more tools for image-based detection of proliferating cells >

Notes:
The Click-iT™ assay can be used on cells in culture or in vivo following EdU administration by feeding or injection methods.
The Click-iT™ assay can be used with BrdU in dual pulse experiments by using the anti-BrdU (clone MoBu-1) antibody, which does not cross react with EdU.
The Click-iT™ technology is compatible with immunohistochemical, immunocytochemical, and fluorescent dyes that are fixation tolerant or designed for fixed cell labeling.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Dye TypeAlexa Fluor™ 594
Emission WavelengthVisible
FormatSlide(s)
Quantity1 kit
Shipping ConditionRoom Temperature
ColorRed
For Use With (Application)Proliferation Assay
For Use With (Equipment)Confocal Microscope, Fluorescence Microscope
Product LineAlexa Fluor, Click-iT, Molecular Probes
Product TypeCell Proliferation Kit
Unit SizeEach
Contents & Storage
Each kit contains enough material for 50 to 250 reactions, depending on reaction volume (typically between 0.1 and 0.5 ml). Store at 2–6°C, dessicate and protect from light.

Frequently asked questions (FAQs)

I will be performing a cell proliferation assay using Click-iT EdU kit. At what point can I stop overnight, or do I have to perform all the steps continuously?

One may store the sample after fixation overnight in PBS at 4oC. For longer storage (<1 week) , store in buffer with 1-2% formaldehyde or in formalin to limit microbial growth. If you use sodium azide as a microbial inhibitor, it must be completely removed prior to the Click-iT reaction.

Find additional tips, troubleshooting help, and resources within our Cell Viability, Proliferation, Cryopreservation, and Apoptosis Support Center.

A control for a Click-iT EdU labeling experiment uses no EdU and the Click-iT reaction using Alexa Fluor 594 azide. The mouse heart tissue sections are showing non-specific labeling in red, seen in particular clusters of cells. They don't overlap with DAPI. What is the problem?

The problem is likely not the Alexa Fluor 594 azide. Since there are no alkynes endogenous to mouse tissue, there is nothing for the dye-azide to bind to. Since the background doesn't overlap with nuclei (DAPI signal), this isn't an issue of unintended EdU labeling. This red is autofluorescence from red blood cells; they autofluoresce in the red and don't have nuclei. This can be confirmed by checking a completely unlabeled tissue section (no dye present at all) to see if they are still present and by examining the cells at high magnification and looking for corpuscular shape.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can I combine Click-iT or Click-iT Plus reactions with phalloidin conjugates used for actin staining?

We do not recommend using phalloidin conjugates for staining actin in combination with traditional Click-iT or Click-iT Plus reactions since phalloidin is extremely sensitive to the presence of copper.

For staining actin in combination with traditional Click-iT or Click-iT Plus reactions, we recommend using anti-α-actin antibodies for staining actin in the cytoskeleton. You can find a list of our actin antibodies here.

Another option would be to use the Click-iT Plus Alexa Fluor Picolyl Azide Toolkit (Cat. Nos. C10641, C10642, C10643). These Click-iT Plus toolkits provide Copper and Copper protectant separately which makes it easier to titrate the copper concentration to obtain optimal labeling with minimal copper-mediated damage. You may need to optimize the click reaction with the lowest possible concentration of copper and then perform the phalloidin staining.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Are the Alexa Fluor azides from Click-iT EdU kits available separately?

Yes, but the standalone products are not shipped at the same amount as provided in the Click-iT EdU kits; the amount of dye-azide provided in the Click-iT kits is proprietary information. See these catalog numbers for the stand alone products:
- Cat. No. A10266: Alexa Fluor 488 azide
- Cat. No. A10270: Alexa Fluor 594 azide
- Cat. No. A10277: Alexa Fluor 647 azide

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I am observing no signal or very low specific signal for my click-labeled samples. What can I do to improve the signal?

The click reaction is only effective when copper is in the appropriate valency. Azides and alkynes will not react with each other without copper. Make sure that the click reaction mixture is used immediately after preparation when the copper (II) concentration is at its highest.
Do not use additive buffer that has turned yellow; it must be colorless to be active.
Cells need to be adequately fixed and permeabilized for the TdT enzyme and click reagents to have access to the nucleus. Tissue samples require digestion with proteinase K or other proteolytic enzymes for sufficient TdT access.
Some reagents can bind copper and reduce its effective concentration available to catalyze the click reaction. Do not include any metal chelator (e.g., EDTA, EGTA, citrate, etc.) in any buffer or reagent prior to the click reaction. Avoid buffers or reagents that include other metal ions that may be o xidized or reduced. It may be help to include extra wash steps on the cell or tissue sample before performing the click reaction.
You can repeat the click reaction with fresh reagents to try to improve signal. Increasing the click reaction time longer than 30 minutes will not improve a low signal. Performing a second, 30 minute incubation with fresh click reaction reagents is more effective at improving labeling.
Your cells may not be apoptotic. Prepare a DNase I-treated positive control to verify that the TdT enzymatic reaction and click labeling reaction are working correctly.

Find additional tips, troubleshooting help, and resources within our Labeling Chemistry Support Center.

Citations & References (32)

Citations & References
Abstract
BRCA1-associated exclusion of 53BP1 from DNA damage sites underlies temporal control of DNA repair.
Authors:Chapman JR, Sossick AJ, Boulton SJ, Jackson SP,
Journal:J Cell Sci
PubMed ID:22553214
'Following irradiation, numerous DNA-damage-responsive proteins rapidly redistribute into microscopically visible subnuclear aggregates, termed ionising-radiation-induced foci (IRIF). How the enrichment of proteins on damaged chromatin actually relates to DNA repair remains unclear. Here, we use super-resolution microscopy to examine the spatial distribution of BRCA1 and 53BP1 proteins within single IRIF at ... More
Quiescent gastric stem cells maintain the adult Drosophila stomach.
Authors:Strand M, Micchelli CA,
Journal:Proc Natl Acad Sci U S A
PubMed ID:21984734
'The adult Drosophila copper cell region or "stomach" is a highly acidic compartment of the midgut with pH < 3. In this region, a specialized group of acid-secreting cells similar to mammalian gastric parietal cells has been identified by a unique ultrastructure and by copper-metallothionein fluorescence. However, the homeostatic mechanism ... More
Long-term live imaging provides new insight into stem cell regulation and germline-soma coordination in the Drosophila ovary.
Authors:Morris LX, Spradling AC,
Journal:Development
PubMed ID:21558370
'The Drosophila ovariole tip produces new ovarian follicles on a 12-hour cycle by controlling niche-based germline and follicle stem cell divisions and nurturing their developing daughters. Static images provide a thumbnail view of folliculogenesis but imperfectly capture the dynamic cellular interactions that underlie follicle production. We describe a live-imaging culture ... More
Histone H3 lysine 4 methylation marks postreplicative human cytomegalovirus chromatin.
Authors:Nitzsche A, Steinhäusser C, Mücke K, Paulus C, Nevels M,
Journal:J Virol
PubMed ID:22761369
'In the nuclei of permissive cells, human cytomegalovirus genomes form nucleosomal structures initially resembling heterochromatin but gradually switching to a euchromatin-like state. This switch is characterized by a decrease in histone H3 K9 methylation and a marked increase in H3 tail acetylation and H3 K4 methylation across the viral genome. ... More
Schwann cell myelination requires integration of laminin activities.
Authors:McKee KK, Yang DH, Patel R, Chen ZL, Strickland S, Takagi J, Sekiguchi K, Yurchenco PD,
Journal:J Cell Sci
PubMed ID:22767514
'Laminins promote early stages of peripheral nerve myelination by assembling basement membranes (BMs) on Schwann cell surfaces, leading to activation of ß1 integrins and other receptors. The BM composition, structural bonds and ligands needed to mediate this process, however, are not well understood. Mice hypomorphic for laminin ?1-subunit expression that ... More