Search
Global leaders in mining exploration and production trust Thermo Scientific solutions and instruments that have been serving the needs of the mining industry for many years.
Whether our customers are improving mineral recovery, assessing critical inputs for metallurgical accounting and particle size analysis, or achieving environmental and ESG compliance throughout the processing value-chain, we are here to support them. At Thermo Fisher Scientific, our solutions include high availability samplers, elemental and particle size analyzers, handheld and laboratory XRF analyzers, bulk weighing and monitoring equipment as well as comprehensive environmental monitoring instruments. Shaped by decades of working with the mining industry, this technology enables optimization of mine life and plant feed grade, yield, efficiency, profitability. Our technology also helps to make mining, greener.
From exploration through processing, from mineral analysis to complete bulk weighing monitoring and sampling systems to informatics, we provide the latest equipment and technology for your mineral and metals operations.
Confidently perform elemental analysis from early stage discovery through mineral processing with the Thermo Scientific handheld XRF analyzer. Our mineral analysers provide miners solutions for rapid geochemical analysis that increase discovery success rates, identify drill targets quickly, make on-site decisions about whether to stop or continue drilling, and decide where to focus on the grid.
CB Omni Agile provides real-time online elemental analysis of an entire raw material process stream using a choice of Prompt Gamma Neutron Activation Analysis (PGNAA) or Pulsed Fast Thermal Neutron Activation (PFTNA) technology. Incorporating a modular industrial design, the analyzer is overall lighter and easier to install than previous generation analyzers. Unprecedented fiexibility allows for the analyzer to be configured by the application and process control needs.
Measure the particle size of concentrated slurries, in real-time, from 1 to 1000 µm, with no dilution and high availability
Sample slurries with a low head, high availability, high accuracy system that offers value-added options for mining, such as pebble screen and distribution.
The Thermo Scientific™ GS Omni elemental analyzer provides accurate, real-time elemental analysis of multiple slurry streams for both light elements. Leveraging the Prompt Gamma Neutron Activation Analysis (PGNAA) technology, the GS Omni elemental analyzer outperforms instruments using X-ray fiuorescence as it directly measures elements lower than calcium on the periodic table. The GS Omni elemental analyzer allows for process control of contaminant elements and minimizes penalty costs as product grades are archieved. It also optimizes reagent dosage and maintains circuit stability.
To ensure accurate process control and optimum productivity, non-contact nuclear density gauges measure the density or percent solids of a liquid or slurry in a pipe. They are used for applications containing a high temperature, highly toxic or highly corrosive process.
The Thermo Scientific™ DDG3 Digital Density Gauge is a high performance, digital, non-intrusive, non-contact nuclear device. It is suitable for applications containing a high temperature, highly toxic or highly corrosive process such as mining, petrochemical, coal, food processing, water treatment, dredging, or pulp and paper.
Combine sampling and XRF analysis in one low head, high availability solution to measure streams from 4 to 50,000 m3/hr in real-time.
Combine multiple streams into one centralized, cost-efficient, low-head, elemental analyzer with no risk of cross-contamination.
Offers the ultimate performance, versatility and reliability of elemental analysis. It is an easy to use and transportable bench-top instrument for rapid screening and finger-printing of ores, minerals, and industrial products for the major and minor elements or oxides.
Solve analytical challenges in a wide range of applications with one comprehensive tool. The Thermo Scientific™ ARL™ PERFORM'X Sequential X-Ray Fluorescence Spectrometer provides perfect solution for geochemical, R&D and contract labs with demanding quantitative mineral analysis
Because mining sites are usually remote, they often utilize coal-fired power plants to supply power to a mining site. To comply with environmental guidelines, emissions should be continuously monitored for toxic fumes and other pollutants. Thermo ScientiFIc Continuous emissions monitoring systems (CEMS) analyser help mining operations monitor SO2 , NOx ,Co, O2 , Hg.
Fugitive dust is typically the result from activities such as the physical movement of soil, vehicles traveling over unpaved surfaces, heavy equipment operation, blasting and wing - all found in mining operations. Thermo Scientific™ ADR™ 1500, real time Ambient Dust Monitor, based on Light scattering technology, is designed for continuous measurement of airborne particles for total particulate and cut-points ranging from PM10 down to PM1. Suitable for fugitive dust monitoring, exposure monitoring, perimeter monitoring, area monitoring and site remediation.
Coal mines must meet a variety of customer specifications for different coal qualities.The Thermo Scientific™ ECA-3 Elemental Crossbelt Analyzer provides real-time quality analysis of critical process streams to facilitate sorting, blending and out-of-seam dilution control.
When real-time knowledge of coal quality is critical, coal-fired power plants and coal producers can rely on the Thermo Scientific™ CQM FLEX (Coal Quality Manager FLEX) Analyzer. The CQM Flex can incorporate an electrically driven neutron generator, or Californium (252Cf) as its source of neutrons to meet the needs of various application requirements.