Search
Adapting a rapid, accurate testing method for detecting contaminants is a key step in enabling in-process testing. The results from those tests can then be used as part of downstream process characterization to make fact-based, go/no-go forward processing decisions.
The AutoMate Express Nucleic Acid Extraction System is an easy-to-use benchtop instrument that enables hands-free automation of the nucleic acid extraction process for contaminant testing. The instrument uses the established PrepSEQ nucleic acid extraction chemistry for quantitative recovery of DNA and RNA from diverse sample types, including cell culture, raw material (i.e., serum), and in-process downstream purification samples such as column purification or bulk drug substance (BDS) samples.
Explore this app note to discover a simplified workflow for simultaneous nucleic acid extraction of mycoplasma, MMV, and vesivirus. This approach, using AutoMate Express with PrepSEQ Express, improves speed, accuracy, and contamination monitoring in bioproduction.
AutoMate Express is a strong fit for labs that want a simplified, cartridge-based automated extraction workflow for contamination testing.
AutoMate Express System uses PrepSEQ Express kits with prefilled cartridges and workflow plastics to automate nucleic acid extraction steps in a closed system.
The workflow is designed to simplify setup, reduce manual handling, and support reproducible extraction from contaminant testing samples.
Workflow steps:
Sealed PrepSEQ Express cartridges help simplify reagent handling and support a contained extraction workflow.
Automated extraction helps reduce hands-on time compared with manual workflows.
AutoMate Express is especially relevant for mycoplasma, MMV, and vesivirus sample preparation workflows using PrepSEQ Express chemistry.
The benchtop system can fit into existing lab workflows where space, simplicity, and ease of operation are important.
Figure 1. DNA recovery from spikes of DNA into various media. Detection of (A) mycoplasma DNA and (B) MMV DNA spiked into PBS (blue), fetal calf serum (black), CHO medium (gray), and conditioned CHO medium (brown). For each matrix, a recovery control (RC) was used to assess extraction efficiency.
Figure 2. DNA recovery from cell culture matrices. Detection of DNA from (A) M. arginini and (B) MMV spiked into PBS (blue), fetal calf serum (black), CHO medium (gray), and CHO conditioned medium (brown). For each plate, control reactions were run with 500, 1,000, and 2,000 copies of the mycoplasma discriminatory positive control or MMV positive control in the real-time PCR assay.
Figure 3. One-hour workflow for the AutoMate Express system.
AutoMate Express and KingFisher systems support different automation needs.
Choose AutoMate Express when you want a closed, cartridge-based workflow using PrepSEQ Express chemistry.
Consider KingFisher systems when you need plate-based magnetic bead automation, higher throughput, or scalable PrepSEQ workflows.
Explore the Analytics Knowledge Hub and discover a robust collection of webinars, articles, eBooks, infographics, and other digital resources to help you optimize analytical testing in your bioprocess workflow.
No matter your questions, we are here to help. Our experienced team of scientists and technical specialists is ready to assist you in choosing the right solution and products. Complete this form to start the conversation.
For Research Use Only. Not for use in diagnostic procedures.