86f4 - Techniques/Products/Contact Us/Learning Center (LS)
Electron Microscopy
Infectious Disease Research using Cryo Electron Microscopy
Cryo electron microscopy techniques for high-resolution structural characterization of infectious diseases.
With an ever-increasing world population and global travel on the rise, infectious diseases cover more ground, spreading faster and more efficiently, leading to an increase in pandemic threats. Besides preventative (quarantine) measures, it is critical to combat diseases directly through the development of new antibiotics and/or efficient vaccines.
Particularly for viruses, determining the 3D outer shell structure can be vital, as that is where neutralizing antibodies interact with the pathogen. Cryo-electron microscopy (cryo-EM) techniques, such as single particle analysis (SPA), cryo electron tomography (cryo-ET), and micro electron diffraction (MicroED), allow multiscale observation of these biological structures in their near-native states. With the full 3D structure, treatments can be motivated by observations on the molecular scale, greatly increasing specificity and overall success.
Thermo Fisher Scientific’s cryo-EM solutions are specially designed to deliver this vital information, leading to faster and more efficient development of therapeutics.
3D SPA reconstruction of the Zika virus and antigen-antibody target sites
3D SPA reconstruction of the Zika virus and antigen-antibody target sites. Image courtesy of Sirohi et al., Purdue University
Samples
Proteins Analysis
Cryo-electron microscopy provides near-atomic resolution 3D protein structure. It can determine structural information for complexes and crystallization-resistant samples, as well as vital cellular context.
Learn more ›
Biopharmaceutical Research
Structural drug discovery is enabled by cryo-electron microscopy, as the method provides near-atomic-resolution detail for small molecules and protein biologics in their fully hydrated state.
Learn more ›
Virus Analysis
Cryo-EM enables the 3D structural visualization of virus particles, and the antigen-antibody interface, at near-atomic resolutions. A virus’s inherent structural symmetry makes it the ideal target for cryo-EM analysis.
Learn more ›
Techniques
Single Particle Analysis
Single particle analysis (SPA) is a cryo-electron microscopy technique that enables structural characterization at near-atomic resolutions, unraveling dynamic biological processes and the structure of biomolecular complexes/assemblies.
Learn more ›
Cryo-Tomography
Cryo-electron tomography (cryo-ET) delivers both structural information about individual proteins as well as their spatial arrangements within the cell. This makes it a truly unique technique and also explains why the method has such an enormous potential for cell biology. Cryo-ET can bridge the gap between light microscopy and near-atomic-resolution techniques like single-particle analysis.
Learn more ›
MicroED
MicroED is an exciting new technique with applications in the structural determination of small molecules and protein. With this method, atomic details can be extracted from individual nanocrystals (<200 nm in size), even in a heterogeneous mixture.
Learn more ›
Single Particle Analysis
Single particle analysis (SPA) is a cryo-electron microscopy technique that enables structural characterization at near-atomic resolutions, unraveling dynamic biological processes and the structure of biomolecular complexes/assemblies.
Learn more ›
Cryo-Tomography
Cryo-electron tomography (cryo-ET) delivers both structural information about individual proteins as well as their spatial arrangements within the cell. This makes it a truly unique technique and also explains why the method has such an enormous potential for cell biology. Cryo-ET can bridge the gap between light microscopy and near-atomic-resolution techniques like single-particle analysis.
Learn more ›
MicroED
MicroED is an exciting new technique with applications in the structural determination of small molecules and protein. With this method, atomic details can be extracted from individual nanocrystals (<200 nm in size), even in a heterogeneous mixture.
Learn more ›
Products
Style Sheet for Instrument Cards Original
Thermo Scientific Krios G4 Cryo transmission electron microscope (TEM)
Krios G4 Cryo-TEM
  • Improved ergonomics
  • Fits more easily into new and existing labs
  • Maximized productivity and automation
  • Best image quality for high-resolution 3D reconstruction
Thermo Scientific Glacios Cryo transmission electron microscope (TEM)
Glacios Cryo-TEM
  • Flexible Accelerating Voltage 80-200 kV
  • Industry-leading Autoloader for cryogenic sample manipulation
  • Small footprint
  • Enhanced ease-of-use
Thermo Scientific Talos Arctia transmission electron microscope (TEM)
Talos Arctica TEM
  • Increased data acquisition speed
  • High data with robotic sample handling & automated loading
  • Unattended platform operation and automated data acquisition
  • Low cost of ownership with remote diagnostics and preventive service
Thermo Scientific Talos F200C transmission electron microscope (TEM)
Talos F200C TEM
  • Flexible EDS analysis reveals chemical information
  • High-contrast, high-quality TEM and STEM imaging
  • Ceta 16 Mpixel CMOS camera provides large field of view and high read-out speed
Thermo Scientific Talos L120C transmission electron microscope (TEM)
Talos L120C TEM
  • Increased stability
  • 4k × 4K Ceta CMOS camera
  • TEM magnification range from 25–650 kX
  • Flexible EDS analysis reveals chemical information
Thermo Scientific Prisma E scanning electron microscope (SEM)
Prisma E SEM
  • Entry-level SEM with excellent image quality
  • Easy and quick sample loading and navigation for multiple samples
  • Compatible with a wide range of materials thanks to dedicated vacuum modes
Thermo Scientific Quattro E scanning electron microscope (SEM)
Quattro ESEM
  • Ultra-versatile high-resolution FEG SEM with unique environmental capability (ESEM)
  • Observe all information from all samples with simultaneous SE and BSE imaging in every mode of operation
Thermo Scientific Apreo 2 scanning electron microscope (SEM)
Apreo 2 SEM
  • High-performance SEM for all-round nanometer or sub-nanometer resolution
  • In-column T1 backscatter detector for sensitive, TV-rate materials contrast
  • Excellent performance at long working distance (10 mm)
Thermo Scientific Verios 5 XHR scanning electron microscope (SEM)
Verios 5 XHR SEM
  • Monochromated SEM for sub-nanometer resolution over the full 1 keV to 30 keV energy range
  • Easy access to beam landing energies as low as 20 eV
  • Excellent stability with piezo stage as standard
VolumeScope 2 SEM
  • Isotropic 3D data from large volumes
  • High contrast and resolution in high and low vacuum modes
  • Simple switch between normal SEM use and serial block-face imaging
Thermo Scientific Helios Hydra plasma focused ion beam scanning electron microscope (DualBeam)
Helios Hydra DualBeam
  • 4 fast switchable ion species (Xe, Ar, O, N) for optimized PFIB processing of a widest range of materials
  • Ga-free TEM sample preparation
  • Extreme high resolution SEM imaging
Thermo Scientific Scios 2 plasma focused ion beam scanning electron microscope (DualBeam)
Scios 2 DualBeam
  • Full support of magnetic and non-conductive samples
  • High throughput subsurface and 3D characterization
  • Advanced ease of use and automation capabilities
Thermo Scientific Aquilos 2 cryo focused ion beam (FIB)
Aquilos 2 Cryo-FIB
  • Automation enables production of multiple lamellas
  • Target and extract your structure of interest with lift-out nano-manipulator
  • 3D visualization for high-resolution tomography
Thermo Scientific Vitrobot vitrification sample system
Vitrobot System
  • Fully automated sample vitrification
  • Blotting device
  • Semi-automated grid transfer
  • High sample throughput
Thermo Scientific Selectris and Selectris X cryo electron microscopy imaging filters
Selectris and Selectris X Imaging Filters
  • Designed for high stability and atomic resolution imaging
  • Straightforward operation
  • Paired with the latest generation Thermo Scientific Falcon 4i Direct Electron Detector
Thermo Scientific Falcon 4i Electron Detector
Falcon 4i Detector
  • High throughput for more images per hour
  • Unsurpassed imaging quality with high DQE
  • Lossless data compression with EER
Thermo Scientific TEM Tomography 5 software
Tomography 5
Software
  • Automated multi-site batch tomography
  • Automatic cassette mapping
  • Dose symmetric tilt scheme for optimal electron dose
  • Fully integrated with Selectris Imaging Filters
Thermo Scientific Maps electron microscopy software
Maps Software
  • Acquire high-resolution images over large areas
  • Easily find regions of interest
  • Automate image acquisition process
  • Correlate data from different sources
Thermo Scientific Auto Slice and View 4.0 serial section electron microscopy software
Auto Slice and View 4.0 Software
  • Automated serial sectioning for DualBeam
  • Multi-modal data acquisition (SEM, EDS, EBSD)
  • On-the-fly editing capabilities
  • Edge based cut placement
Thermo Scientific Inspect 3D tomography software
Inspect 3D Software
  • Image processing tools and filters for cross-correlation
  • Feature tracking for image alignment
  • Algebraic reconstruction technique for iterative projection comparison
Thermo Scientific Amira 2D-5D visualization Software
Amira Software
Life Sciences
  • Explore 2D-5D bioimaging data
  • Identify and understand structures
  • Obtain statistical information
  • Share reports and stunning animations
Thermo Scientific EPU Software for single particle analysis
EPU 2 Software
  • Microscope-embedded solution for single particle acquisition
  • Optimized for high-throughput particle collection
  • Compatible with film, CCD cameras, and direct electron detectors
Contact us
Style Sheet for Komodo Tabs
Style Sheet to change Applications H3 to p with em-h3-header class
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards