Front right angled view of Countess 3 instrument with 2 light cubes at the left side
Accurate cell viability data in less than 30 seconds
The Countess 3 FL Automated Cell Counter has fluorescence capabilities with two fluorescence channels that can easily be changed with 15 light cubes to choose from, offering class-leading flexibility. With the option for fluorescence capabilities, the Invitrogen Countess 3 FL Automated Cell Counter can count cells in brightfield, while also reporting and measuring fluorescent protein expression, apoptosis, and cell viability using fluorescent reagents in as little as 30 seconds. Fluorescence cell counting should be considered when using fluorescence cellular assays that will provide more specificity to complex samples and/or sensitivity for low-abundance or hard-to-detect cells.
The Countess 3 FL Automated Cell Counter is a highly accurate brightfield and dual fluorescence cell counter, with interchangeable light cubes.
Fluorescence capabilities—select any two fluorescence colors from 15 light cube options
The Countess 3 FL instrument has two optional fluorescence channels. The fluorescent colors collected are determined by the insertion of individual light cubes. With 15 light cubes to choose from, the Countess 3 FL instrument is not limited to detection of GFP- and RFP-expressing cells. Interested in measuring apoptosis and viability of cells in a population? No problem. Use an apoptosis indicator that measures caspase activation, such as Invitrogen CellEvent Caspase-3/7 Green Detection Reagent along with the EVOS GFP Light Cube. Additionally, use a viability dye such as Invitrogen SYTOX Red Dead Cell Stain with the EVOS Cy5 Light Cube to measure dead cells that have progressed past apoptosis. This staining combination along with the two EVOS light cubes offers a fast and simple method to obtain apoptosis and viability data.
Seven EVOS light cubes lined up
Cell viability assessment
Assessing cell viability is a key step in daily cell culture and is required for accurate and efficient downstream processing. With the Countess 3 FL instrument, there are fluorescence options that offer a sensitive and specific determination of viability. These can be done quickly and easily to check viability prior to downstream sample analysis. By using Invitrogen ReadyCount Green/Red Viability Stain which contains acridine orange and propidium iodide (AO/PI) stains in combination, the fluorescence staining provides great sensitivity and greater specificity of the live/dead status amid a mixed cell population, and it does not alter the morphology of the cell (Figure 1).
Measure transfection efficiency
The ability to assess the number of cells successfully transfected or transduced in a cell population is a basic and critical evaluation parameter. Commonly, transfection or transduction of cells of interest results in the expression of a fluorescent protein reporter, such as GFP. The Countess 3 FL Automated Cell Counter, loaded with your choice of light cubes, offers a quick and simple method to easily obtain transfection efficiency data (Figure 2).
Figure 2. Determine the percentage transfection efficiency using the Countess 3 FL Automated Cell Counter. Cells can be seen adhered to the vessel and expressing GFP fluorescence on the EVOS M5000 Imaging System (A). Using the Countess 3 Automated Cell Counter, trypsinized cells can be counted and transfection efficiency can be quantified (B).
Countess 3 Automated Cell Counter
Watch a brief video with some of the many features developed for the Countess 3 instruments.
How to add and change Invitrogen EVOS Light Cubes
Add or change light cubes on the Countess 3 FL Automated Cell Counter, allowing flexibility and customization based on assay requirements.
How to perform fluorescence cell counting and analysis using layers & contours
Use fluorescence cell assays to perform cell counts for deeper analysis.
How to visualize histograms and perform gating
Evaluate fluorescence images using the histogram and gating features.
How to use the reusable slide and accelerate your cell counting with Rapid capture
The reusable slide is a cost effective way to perform automated cell counting and Rapid capture will minimize steps when using the Countess 3 Automated Cell Counter in brightfield mode.
How to save your data
Following cell counts, data can be saved in the cloud or to a USB drive. Select from five file types and select Autosave to automatically save the files of your choice.
How to use onboard calculators
Use the pre-dilution calculator to correct for dilutions prior to counting including a quick trypan blue 1:1 dilution correction selection. Use the cell splitting calculator to set up cell culture vessels with the optimal cell densities.
The result, image, and data files can be easily saved to a USB drive or by Wi-Fi enabled connectivity to the Cloud using Thermo Fisher Connect. There are five different files that can be saved. Autosave enables automatic saving of data, so there is no need to select Save after each count.
  • The “Result” and “Images” files can be saved as JPEG, TIFF, or PNG files.
  • The “Report” file is a PDF file and includes the result and image files.
  • Data files are in CSV format, which:
    • can be easily imported into your current analysis software or viewed in Excel software.
    • has the option to save All, Last 30 Days, Last 7 Days, Today, or Current.
  • The “Cell Data” file is an FCS file for advanced individual cell data plotting and analysis.
Save options screen shows various files that can be saved, data destinations, and editable field for sample name along with field for enabling autosave.
Counting has been successfully performed on the following cell lines using the Countess 3 FL Automated Cell Counter:
Cell Animal Organ
HeLa Human Cervix
U2OS Human Bone
A549 Human Lung
CHO K1 Hamster Ovary
HEK293 Human Kidney
SKBR-3 Human Breast
BPAE Bovine Smooth muscle
HCASM Human Smooth muscle
Jurkat Human Blood
Ramos Human Blood
THP-1 Human Macrophage
RAW Mouse Macrophage
MMM Mouse Macrophage
U937 Human Blood
HepG2 Human Liver
MCF-7 Human Breast
Human PBMC Human Blood
Mouse PBMC Mouse Blood
Mouse Hepatocyte Mouse Liver
CAR-T Human Blood
Human Hepatocyte Human Liver
Rat Hepatocyte Rat Liver
Human Spleenocyte Human Spleen
Technical specifications
  • Processing time: <30 seconds
  • Sample concentration range: 1 x 104–1 x 107 cells/mL
  • Particle diameter range: ~4–60 µm
  • Cell diameter range: ~4–60 µm
  • Required sample volume: 10 μL
  • USB drive: 32 GB (FAT32-, exFAT-, or NTSF-compatible)
  • Optics: 5 megapixel camera, 2.5X optical magnification
Physical characteristics
  • Instrument dimensions:
    9" (W) x 5½" (D) x 9" (H); 228.6 mm (W) X 139.7 mm (D) x 228.6 mm (H)
  • Weight: 8 lbs
Counting area
  • Area: 2.26 mm x 1.69 mm (3.82 mm2)
    Graphic image depicting the area of a typical standard hemocytometer an and the area counted of by the Countess 3 Cell counter which is significantly larger
Included with instrument
  • USB drive for storing and transferring data to your computer
  • Disposable Slide Holder
  • Reusable Slide Holder
  • Countess Cell Counting Chamber Slides (Qty 50)
  • Wi-Fi Dongle
  • Power cord and 4 plug adaptors to accommodate most countries
  • Quick reference card (QRC)
Sold separately
The Countess Automated Cell Counter Systems supported with:
  • 3 year extended warranty packages covering all costs for shipping, parts, and labor for repairs
  • Warranty period access to over 1,400 trained professionals, ready to assist you and help keep your instruments up and running
Keep your Countess System up and running with our instrument service plans, tailored to fit your lab’s needs and budgets:
  Rapid Exchange AB Repair Center (ABRC) Support
Plan is designed for Labs that operate under critical timelines Regulated labs that must maintain asset tagging consistency to comply with regulated protocols
Repair time Replacement shipped out in 1 business day Your instrument repaired and returned in ~3 weeks
Off-site repair service including shipping, parts, and labor    
High-priority telephone and email access to our remote service center    
Visit our Countess Automated Cell Counter Service Plans page to learn more about services and support for your Countess Systems.
Countess 3 Automated Cell Counter
Watch a brief video with some of the many features developed for the Countess 3 instruments.
How to add and change Invitrogen EVOS Light Cubes
Add or change light cubes on the Countess 3 FL Automated Cell Counter, allowing flexibility and customization based on assay requirements.
How to perform fluorescence cell counting and analysis using layers & contours
Use fluorescence cell assays to perform cell counts for deeper analysis.
How to visualize histograms and perform gating
Evaluate fluorescence images using the histogram and gating features.
How to use the reusable slide and accelerate your cell counting with Rapid capture
The reusable slide is a cost effective way to perform automated cell counting and Rapid capture will minimize steps when using the Countess 3 Automated Cell Counter in brightfield mode.
How to save your data
Following cell counts, data can be saved in the cloud or to a USB drive. Select from five file types and select Autosave to automatically save the files of your choice.
How to use onboard calculators
Use the pre-dilution calculator to correct for dilutions prior to counting including a quick trypan blue 1:1 dilution correction selection. Use the cell splitting calculator to set up cell culture vessels with the optimal cell densities.
The result, image, and data files can be easily saved to a USB drive or by Wi-Fi enabled connectivity to the Cloud using Thermo Fisher Connect. There are five different files that can be saved. Autosave enables automatic saving of data, so there is no need to select Save after each count.
  • The “Result” and “Images” files can be saved as JPEG, TIFF, or PNG files.
  • The “Report” file is a PDF file and includes the result and image files.
  • Data files are in CSV format, which:
    • can be easily imported into your current analysis software or viewed in Excel software.
    • has the option to save All, Last 30 Days, Last 7 Days, Today, or Current.
  • The “Cell Data” file is an FCS file for advanced individual cell data plotting and analysis.
Save options screen shows various files that can be saved, data destinations, and editable field for sample name along with field for enabling autosave.
Counting has been successfully performed on the following cell lines using the Countess 3 FL Automated Cell Counter:
Cell Animal Organ
HeLa Human Cervix
U2OS Human Bone
A549 Human Lung
CHO K1 Hamster Ovary
HEK293 Human Kidney
SKBR-3 Human Breast
BPAE Bovine Smooth muscle
HCASM Human Smooth muscle
Jurkat Human Blood
Ramos Human Blood
THP-1 Human Macrophage
RAW Mouse Macrophage
MMM Mouse Macrophage
U937 Human Blood
HepG2 Human Liver
MCF-7 Human Breast
Human PBMC Human Blood
Mouse PBMC Mouse Blood
Mouse Hepatocyte Mouse Liver
CAR-T Human Blood
Human Hepatocyte Human Liver
Rat Hepatocyte Rat Liver
Human Spleenocyte Human Spleen
Technical specifications
  • Processing time: <30 seconds
  • Sample concentration range: 1 x 104–1 x 107 cells/mL
  • Particle diameter range: ~4–60 µm
  • Cell diameter range: ~4–60 µm
  • Required sample volume: 10 μL
  • USB drive: 32 GB (FAT32-, exFAT-, or NTSF-compatible)
  • Optics: 5 megapixel camera, 2.5X optical magnification
Physical characteristics
  • Instrument dimensions:
    9" (W) x 5½" (D) x 9" (H); 228.6 mm (W) X 139.7 mm (D) x 228.6 mm (H)
  • Weight: 8 lbs
Counting area
  • Area: 2.26 mm x 1.69 mm (3.82 mm2)
    Graphic image depicting the area of a typical standard hemocytometer an and the area counted of by the Countess 3 Cell counter which is significantly larger
Included with instrument
  • USB drive for storing and transferring data to your computer
  • Disposable Slide Holder
  • Reusable Slide Holder
  • Countess Cell Counting Chamber Slides (Qty 50)
  • Wi-Fi Dongle
  • Power cord and 4 plug adaptors to accommodate most countries
  • Quick reference card (QRC)
Sold separately
The Countess Automated Cell Counter Systems supported with:
  • 3 year extended warranty packages covering all costs for shipping, parts, and labor for repairs
  • Warranty period access to over 1,400 trained professionals, ready to assist you and help keep your instruments up and running
Keep your Countess System up and running with our instrument service plans, tailored to fit your lab’s needs and budgets:
  Rapid Exchange AB Repair Center (ABRC) Support
Plan is designed for Labs that operate under critical timelines Regulated labs that must maintain asset tagging consistency to comply with regulated protocols
Repair time Replacement shipped out in 1 business day Your instrument repaired and returned in ~3 weeks
Off-site repair service including shipping, parts, and labor    
High-priority telephone and email access to our remote service center    
Visit our Countess Automated Cell Counter Service Plans page to learn more about services and support for your Countess Systems.
What are other great benefits of automated cell counting?
  • Accuracya highly accurate machine-learning algorithm is applied to cell counting.
  • Precisionauto-lighting and autofocus minimize user inputs and variability.
  • SpeedRapid capture and autosave features generate and save results faster.
  • Convenientbuilt-in pre-dilution and cell splitting calculators, histograms and gating, and a results PDF report.
  • Affordable—both Countess 3 models are compatible with the Countess Reusable Slide.
Learn more about automated cell counters
Countess 3 FL Automated Cell Counter ordering information
Countess 3 FL Automated Cell Counter Productivity Package
A custom quote will be provided after selecting from the following items:
  • Countess 3 FL Automated Cell Counter
  • Countess 3 FL Reusable Slide
  • Countess Cell Counting Chamber Slides (Qty 50)
  • Countess Cell Counting Chamber Slides (Qty 500)
  • Invitrogen Trypan Blue Stain (0.4%)
  • EVOS Light Cube, GFP 2.0
  • EVOS Light Cube, RFP 2.0
  • EVOS Light Cube, DAPI 2.0
  • ReadyCount Green/Red Viability Stain
  • 3 years Rapid Exchange Extended Warranty
Request a quote