Benchtop Cooler
The Nalgene™ Quick Chill™ Unit is designed to rapidly and repeatedly cool down biological samples such as ethanol precipitations for DNA/RNA, bacteria and viruses.
  • Rapidly and repeatedly chills biological samples up to -20°C
  • Can be used repeatedly for up to 45 minutes eliminating the inconvenience of dry ice/alcohol baths
  • Break-resistant polycarbonate stackable design with a convenient carrying handle
  • Rubber feet to keep the unit from sliding on the bench surface
The self-contained unit eliminates the need to use traditional coolants such as dry ice/alcohol or acetone slurries which can be difficult to obtain and unsafe to handle. There is no contact with the coolant so the label or marking stays intact.
Available Thermo Scientific Benchtop Coolers
-20˚C Compatible Benchtop Coolers
Cat.
#355501
Cat. #5115-0012
Cat. #5115-0032
Cat. #DS5114-0012
# of Compartments
4 x 5 (20 total) 2 x 4 (12 total) 4 x 8 (32 total) 3 x 4 (12 total)
Tube Compatibility 0.2 - 0.5mL
0.2 - 0.5mL
0.5 - 0.5mL Cryovials 0.2 - 1.5mL
Style Non-filled / Clear lid Non-filled / Clear lid Gel-filled / White lid Gel-filled / Clear lid
Holding Temp. <-15°C, 1 hr <-15°C, 1 hr <-15°C, 2 hrs <-15°C, 45 mins
Temp Rating
-20°C -20°C -20°C -20°C
  Order now Order now Order now Order now
0˚C Compatible Benchtop Coolers
Cat. #5116-1300
Cat. #5116-1600
Cat. #DS5116-0012
Cat. #DS5116-0032
# of Compartments
3 x 4 (12 total) 3 x 4 (12 total) 3 x 4 (12 total) 4 x 8 (32 total)
Tube Compatibility 12 - 13mm diameter tubes
16 - 17mm diameter tubes
0.2 - 0.5mL Cryovials and microcentrifuge tubes 0.2 - 0.5mL Cryovials and microcentrifuge tubes
Style Non-filled / Clear lid Non-filled / Clear lid Non-filled / Clear lid Gel-filled / White lid
Holding Temp. <1°C, 5.0 hrs <1°C, 5.0 hrs <1°C, 3.5 hrs <1°C, 5.0 hrs
Temp Rating
0°C 0°C 0°C 0°C
  Order now Order now Order now Order now
-20˚C Compatible Benchtop Coolers
Cat.
#355501
Cat. #5115-0012
Cat. #5115-0032
Cat. #DS5114-0012
# of Compartments
4 x 5 (20 total) 2 x 4 (12 total) 4 x 8 (32 total) 3 x 4 (12 total)
Tube Compatibility 0.2 - 0.5mL
0.2 - 0.5mL
0.5 - 0.5mL Cryovials 0.2 - 1.5mL
Style Non-filled / Clear lid Non-filled / Clear lid Gel-filled / White lid Gel-filled / Clear lid
Holding Temp. <-15°C, 1 hr <-15°C, 1 hr <-15°C, 2 hrs <-15°C, 45 mins
Temp Rating
-20°C -20°C -20°C -20°C
  Order now Order now Order now Order now
0˚C Compatible Benchtop Coolers
Cat. #5116-1300
Cat. #5116-1600
Cat. #DS5116-0012
Cat. #DS5116-0032
# of Compartments
3 x 4 (12 total) 3 x 4 (12 total) 3 x 4 (12 total) 4 x 8 (32 total)
Tube Compatibility 12 - 13mm diameter tubes
16 - 17mm diameter tubes
0.2 - 0.5mL Cryovials and microcentrifuge tubes 0.2 - 0.5mL Cryovials and microcentrifuge tubes
Style Non-filled / Clear lid Non-filled / Clear lid Non-filled / Clear lid Gel-filled / White lid
Holding Temp. <1°C, 5.0 hrs <1°C, 5.0 hrs <1°C, 3.5 hrs <1°C, 5.0 hrs
Temp Rating
0°C 0°C 0°C 0°C
  Order now Order now Order now Order now
Support
Need technical support? Contact our expert team for technical and application support of Laboratory Products