Cryogenic Markets
Cryogenic storage is a harsh environment that can shrink or peel box and tube labels and cause writing to deteriorate and become unreadable over time. It's important to use cryogenically compatible markers, labels, and pens to ensure you will always be able to track and locate your precious samples when you need them.
Cryo Markers and Pens
Label cryogenic vials to -200°C (-328°F) with the Thermo Scientific™ Cryo Marker Pen Set. The ink is fast-drying, nonsmearing and permanent.
Catalog #DescriptionColorQuantity
6313-0010Nalgene Cryoware Marker SetAssorted (Red, Blue, Green, Black)24
6313-0020Nalgene Cryoware Marker SetBlack24
343850Nunc Cryo Pen Permanent MarkerBlack10
Cryoware Labels
25 x 50mm cloth labels specifically designed for use at ultra-low temperatures. These labels will accept markers or ballpoint pen and adhere to plastic and cardboard without shrinking or peeling.
Support
Need technical support? Contact our expert team for technical and application support of Laboratory Products