Search
C57BL/6 mouse bone marrow cells were stained with CD45 Monoclonal Antibody, FITC (Product ## 11-0451) and 0.25 µg of Rat IgG2b kappa Isotype Control, Super Bright 600 (Product ## 63-4031) (left) or 0.25 µg of TER-119 Monoclonal Antibody, Super Bright 600 (Product ## 63-5921) (right). Total viable cells were used for analysis.