    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • VGLUT1 Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          Invitrogen
          VGLUT1 Polyclonal Antibody
          Advanced Verification
          1 Published Figure
          2 References
          View all (49) VGLUT1 antibodies
          Protocols
          Cite VGLUT1 Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          • Published Figures (1)
          • Advanced Verification (1)
          VGLUT1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          VGLUT1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          VGLUT1 Antibody (OSV00007G) in WB
          Western blot was performed using Anti-VGLUT1 Monoclonal Antibody (Product # OSV00007G) and a 50 kDa band corresponding to VGLUT1 was observed across tissues tested Mouse Brain (Lane 1), Rat Brain (Lane 2) and Mouse Spleen (Lane 3) were electrophoresed using NuPAGE™ 4-12% Bis-Tris Protein Gel (Product # NP0322BOX). Resolved proteins were then transferred onto a nitrocellulose membrane (Product # IB23001) by iBlot® 2 Dry Blotting System (Product # IB21001). The blot was probed with the primary antibody (4µg/ml) and detected by chemiluminesce... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          VGLUT1 Antibody in Western Blot (WB)
          VGLUT1 Antibody in Immunocytochemistry (ICC/IF)
          VGLUT1 Antibody
          Product Details
          OSV00007G
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          4-50 µg/mL
          -
          Immunohistochemistry (IHC)
          10-50 µg/mL
          -
          Immunocytochemistry (ICC/IF)
          -
          View 2 publications 2 publications
          Flow Cytometry (Flow)
          10-50 µg/mL
          -
          Product Specifications
          Published species
          Human
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          A synthetic peptide from rat VGluT1 conjugated to blue carrier protein was used as the antigen.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Lyophilized
          Concentration
          1 mg/mL
          Amount
          500 µg
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Glycerol (1:1) may be added for added stability.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_10980943
          Product Specific Information
          OSV00007G is specific for VGLUT1 (both isoforms 1 and 2).
          Reconstitute with 500 µL of distilled water.
          Target Information
          A Na(+)-dependent inorganic phosphate cotransporter; may be involved in neuronal Na+ transport.
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-6b469b8bb7-kg5vd:80/100.66.78.64:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0