  • Applications
    • Real-Time PCR
    • Cloning
    • Protein Biology
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • Carboxypeptidase A1 Antibodies
          Rockland
          Carboxypeptidase A Polyclonal Antibody
          View all (65) Carboxypeptidase A1 antibodies
          Tech Support
          Cite Carboxypeptidase A Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          Carboxypeptidase A Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Carboxypeptidase A Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Carboxypeptidase A Antibody (200-4152S) in WB
          Western Blot of Rabbit Anti-Carboxypeptidase A primary antibody. Lane 1: Carboxypeptidase A. Lane 2: None. Load: 50 ng per lane. Primary antibody: Carboxypeptidase A primary antibody at 1:1,000 overnight at 4°C. Secondary antibody: Peroxidase rabbit secondary antibody at 1:40,000 for 30 min at RT. Blocking: MB-070 for 30 min at RT. Predicted/Observed size: 34 kDa, 34 kDa for Carboxypeptidase A. Other band(s): Carboxypeptidase A splice variants and isoforms. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Carboxypeptidase A Antibody in Western Blot (WB)
          Carboxypeptidase A Polyclonal Antibody
          Product Details
          200-4152S
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000
          -
          Immunohistochemistry (IHC)
          Assay-dependent
          -
          ELISA (ELISA)
          1:250,000
          -
          Immunoprecipitation (IP)
          1:500-1:3,000
          -
          Product Specifications
          Species Reactivity
          Bovine
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Carboxypeptidase A [Bovine Pancreas]
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Lyophilized
          Concentration
          1 mg/mL
          Amount
          25 µg
          Purification
          Ion-exchange chromatography
          Storage buffer
          0.02M potassium phosphate, pH 7.2, with 0.15M NaCl
          Contains
          0.01% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Store vial at 4° C prior to restoration. For extended storage aliquot contents and freeze at -20° C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
          Anti-CARBOXYPEPTIDASE A is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Rabbit Serum as well as purified and partially purified Carboxypeptidase A [Bovine Pancreas]. Cross reactivity against Carboxypeptidase A from other tissues and species may occur but have not been specifically determined.
          Target Information
          CPA1, also known as CPA, is a 419 amino acid secreted monomeric protein that is highly expressed in pancreatic tissue. Functioning as a pancreatic exopeptidase, CPA1 uses zinc as a cofactor to catalyze the release of C-terminal amino acids from a variety of proteins, thereby playing a key role in protein digestion and degradation. Via its catalytic activity, CPA1 is also thought to be involved in zymogen (proenzyme) inhibition, probably functioning to block enzyme activation pathways. Abnormal levels of CPA1 are associated with pancreatic cancer, suggesting a possible role in either tumor progression or tumor suppression events.
          Bioinformatics
          Protein Aliases: Carboxypeptidase A1; carboxypeptidase A1 (pancreatic); pancreatic procarboxypeptidase A
          View more View less
          Gene Aliases: CPA; CPA1
          View more View less
          UniProt ID: (Bovine) P00730
          View more View less
          Entrez Gene ID: (Bovine) 286762
          View more View less
          Function(s)
          metallocarboxypeptidase activity zinc ion binding metalloprotease
          Process(es)
          proteolysis leukotriene metabolic process
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          • Documents and Certificates
          • Study information disclosure
          • Promotions
          • Events and Seminars
          • Blog
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          Japan flag icon
          Japan
          Your items have has been added!
          Host server : magellan-search-green-6479c7f679-mppzp:80/100.66.76.236:80.
          git-commit: b70c0dceb888abc3b0dcebb5bc3a5b8753e7e816
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.50.0-Offline