          • Primary Antibodies ›
          • SVOP Antibodies
          Antibodies Inc
          SVOP Monoclonal Antibody (N356/23), FL550
          View all (7) SVOP antibodies
          Tech Support
          Cite SVOP Monoclonal Antibody (N356/23), FL550
          Additional Information:
          {{banner.description}}
          SVOP Monoclonal Antibody (N356/23), FL550
          Product Details
          75-353-FL550
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N356/23
          Immunogen
          Fusion protein amino acids 1-85 of rat SVOP (accession number Q9Z2I7) produced recombinantly in E. Coli.
          View immunogen
          Conjugate
          FL550 FL550 FL550
          • Unconjugated
          • FL490
          • FL594
          • FL650
          • Request custom conjugation
          Excitation/Emission Max
          550/575 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Sequence of this protein is as follows: MEEDLFQLRQ LPVVKFRRTG ESARSEDDAA SGEHDVQIEG VRVGLEAVEL DDGAAVPKEF ANPTDDTFMV EDAVEAIGFG RFQWK
          Target Information
          SVOP gene ontology (GO) annotation includes transmembrane transporter activity and synaptic vesicle membrane.
          Bioinformatics
          Protein Aliases: SV2-related protein; Synaptic vesicle 2-related protein
          View more View less
          Gene Aliases: 1110030H18Rik; AI415691; msvop; Svop
          View more View less
          UniProt ID: (Rat) Q9Z2I7, (Mouse) Q8BFT9
          View more View less
          Entrez Gene ID: (Rat) 171442, (Mouse) 68666
          View more View less
          Function(s)
          ion transmembrane transporter activity transmembrane transporter activity
          Process(es)
          ion transmembrane transport transmembrane transport transport
          Pedidos Plus Icon Minus Icon
          • Status do pedido
          • Ajuda para pedidos
          • Pedido rápido
          • Supply Center
          • eProcurement
          Suporte Plus Icon Minus Icon
          • Ajuda e suporte
          • Entre em Contato
          • Centros de Suporte Técnico
          • Obter Documentos e Certificados
          • Informe um Problema no Site
          Recursos Plus Icon Minus Icon
          • Centros de aprendizagem
          • Promoções
          • Eventos & Webinars
          • Mídia Sociais
          Sobre a Thermo Fisher Plus Icon Minus Icon
          • Sobre Nós Sobre Nós
          • Carreiras Carreiras
          • Investidores Investidores
          • Sala de Impresa Sala de Impresa
          • Responsabilidade Social Responsabilidade Social
          • Marcas
          • Políticas e avisos
          Nosso Portfólio Plus Icon Minus Icon
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline