    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
          • Primary Antibodies ›
          • SVOP Antibodies
          Antibodies Inc
          SVOP Monoclonal Antibody (N356/23), FL594
          View all (7) SVOP antibodies
          Tech Support
          Cite SVOP Monoclonal Antibody (N356/23), FL594
          Additional Information:
          {{banner.description}}
          SVOP Monoclonal Antibody (N356/23), FL594
          Product Details
          75-353-FL594
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N356/23
          Immunogen
          Fusion protein amino acids 1-85 of rat SVOP (accession number Q9Z2I7) produced recombinantly in E. Coli.
          View immunogen
          Conjugate
          FL594 FL594 FL594
          • Unconjugated
          • FL490
          • FL550
          • FL650
          • Request custom conjugation
          Excitation/Emission Max
          594/615 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Sequence of this protein is as follows: MEEDLFQLRQ LPVVKFRRTG ESARSEDDAA SGEHDVQIEG VRVGLEAVEL DDGAAVPKEF ANPTDDTFMV EDAVEAIGFG RFQWK
          Target Information
          SVOP gene ontology (GO) annotation includes transmembrane transporter activity and synaptic vesicle membrane.
          Bioinformatics
          Protein Aliases: SV2-related protein; Synaptic vesicle 2-related protein
          View more View less
          Gene Aliases: 1110030H18Rik; AI415691; msvop; Svop
          View more View less
          UniProt ID: (Rat) Q9Z2I7, (Mouse) Q8BFT9
          View more View less
          Entrez Gene ID: (Rat) 171442, (Mouse) 68666
          View more View less
          Function(s)
          ion transmembrane transporter activity transmembrane transporter activity
          Process(es)
          ion transmembrane transport transmembrane transport transport
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          India flag icon
          India
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline