          • Primary Antibodies ›
          • SVOP Antibodies
          Antibodies Inc
          SVOP Monoclonal Antibody (N356/23)
          View all (7) SVOP antibodies
          Tech Support
          Cite SVOP Monoclonal Antibody (N356/23)
          Additional Information:
          {{banner.description}}
          SVOP Monoclonal Antibody (N356/23)
          Product Details
          75-353
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N356/23
          Immunogen
          Fusion protein amino acids 1-85 of rat SVOP (accession number Q9Z2I7) produced recombinantly in E. Coli.
          View immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • FL490
          • FL550
          • FL594
          • FL650
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          TBS, pH 7.4
          Contains
          0.065% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Sequence of this protein is as follows: MEEDLFQLRQ LPVVKFRRTG ESARSEDDAA SGEHDVQIEG VRVGLEAVEL DDGAAVPKEF ANPTDDTFMV EDAVEAIGFG RFQWK
          Target Information
          SVOP gene ontology (GO) annotation includes transmembrane transporter activity and synaptic vesicle membrane.
          Bioinformatics
          Protein Aliases: SV2-related protein; Synaptic vesicle 2-related protein
          View more View less
          Gene Aliases: 1110030H18Rik; AI415691; msvop; Svop
          View more View less
          UniProt ID: (Rat) Q9Z2I7, (Mouse) Q8BFT9
          View more View less
          Entrez Gene ID: (Rat) 171442, (Mouse) 68666
          View more View less
          Function(s)
          ion transmembrane transporter activity transmembrane transporter activity
          Process(es)
          ion transmembrane transport transmembrane transport transport
          Contact tech support
          オンラインオーダー Plus Icon Minus Icon
          • オンラインオーダー可能な製品
          • オンラインオーダーに関する個人情報の取扱いについて
          • 特定商取引法に基づく表記
          • 返品について
          • 表示価格について
          • 配送料について
          サポート Plus Icon Minus Icon
          • FAQ、サポート情報
          • お問い合わせ一覧
          • サポートセンター
          • ドキュメント、証明書
          • 研究情報公開
          • Webサイトの問題を報告​
          リソース Plus Icon Minus Icon
          • ラーニングセンター
          • キャンペーン
          • イベント、セミナー
          • Blog
          • 企業情報
          • Social Responsibility (CSR)
          • お客さま体験の向上
          • 採用情報 採用情報
          • ニュース
          • 商標
          • ポリシーと通知
          • PPD
          • 標準販売条件
          • 個人情報保護方針
          Japan flag icon
          Japan
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline