  • Applications
    • Real-Time PCR
    • Cloning
    • Protein Biology
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • SVOP Antibodies
          Antibodies Inc
          SVOP Monoclonal Antibody (N356/23), FL650
          View all (7) SVOP antibodies
          Tech Support
          Cite SVOP Monoclonal Antibody (N356/23), FL650
          Additional Information:
          {{banner.description}}
          SVOP Monoclonal Antibody (N356/23), FL650
          Product Details
          75-353-FL650
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N356/23
          Immunogen
          Fusion protein amino acids 1-85 of rat SVOP (accession number Q9Z2I7) produced recombinantly in E. Coli.
          View immunogen
          Conjugate
          FL650 FL650 FL650
          • Unconjugated
          • FL490
          • FL550
          • FL594
          • Request custom conjugation
          Excitation/Emission Max
          655/676 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Sequence of this protein is as follows: MEEDLFQLRQ LPVVKFRRTG ESARSEDDAA SGEHDVQIEG VRVGLEAVEL DDGAAVPKEF ANPTDDTFMV EDAVEAIGFG RFQWK
          Target Information
          SVOP gene ontology (GO) annotation includes transmembrane transporter activity and synaptic vesicle membrane.
          Bioinformatics
          Protein Aliases: SV2-related protein; Synaptic vesicle 2-related protein
          View more View less
          Gene Aliases: 1110030H18Rik; AI415691; msvop; Svop
          View more View less
          UniProt ID: (Rat) Q9Z2I7, (Mouse) Q8BFT9
          View more View less
          Entrez Gene ID: (Rat) 171442, (Mouse) 68666
          View more View less
          Function(s)
          ion transmembrane transporter activity transmembrane transporter activity
          Process(es)
          ion transmembrane transport transmembrane transport transport
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          • Documents and Certificates
          • Study information disclosure
          • Promotions
          • Events and Seminars
          • Blog
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          Japan flag icon
          Japan
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline