  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • VGAT Antibodies
          Antibodies Inc
          VGAT Monoclonal Antibody (L118/80), FL594
          View all (28) VGAT antibodies
          Tech Support
          Cite VGAT Monoclonal Antibody (L118/80), FL594
          Additional Information:
          {{banner.description}}
          VGAT Monoclonal Antibody (L118/80), FL594
          Product Details
          75-457-FL594
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Mouse, Rat
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          L118/80
          Immunogen
          Fusion protein amino acids 1-133 (cytoplasmic N-terminus) of mouse VGAT (accession number O35633) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          FL594 FL594 FL594
          • Unconjugated
          • FL490
          • FL550
          • FL650
          • Request custom conjugation
          Excitation/Emission Max
          594/615 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          The Vesicular GABA Amino Acid Transporter (VGAT) is responsible for transport of the inhibitory neurotransmitter into synaptic vesicles (McIntire et al., 1997). The VGAT protein (also known as the Vesicular Inhibitory Amino Aid Transporter or VIAAT) is expressed in synaptic vesicles of both glycine and GABAergic synapses throughout the CNS (Chaudhry et al., 1998). Expression of the VGAT protein changes during development and also in response to patterns of neuronal activity (De et al., 2005).
          Bioinformatics
          Protein Aliases: GABA and glycine transporter; GVAT; mVGAT; mVIAAT; rat UNC-47 homolog; rGVAT; RUNC-47; Solute carrier family 32 member 1; vasicular inhibitory amino acid transporter 10D; Vesicular GABA and glycine transporter; Vesicular GABA transporter; Vesicular inhibitory amino acid transporter
          View more View less
          Gene Aliases: R75019; Slc32a1; VGAT; Viaat
          View more View less
          UniProt ID: (Rat) O35458, (Mouse) O35633
          View more View less
          Entrez Gene ID: (Rat) 83612, (Mouse) 22348
          View more View less
          Function(s)
          amino acid transmembrane transporter activity glycine transmembrane transporter activity gamma-aminobutyric acid:proton symporter activity
          Process(es)
          amino acid transmembrane transport neurotransmitter transport aging gamma-aminobutyric acid transport glycine transport proton transport sodium ion transmembrane transport L-alpha-amino acid transmembrane transport transport
          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          Taiwan flag icon
          Taiwan
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline