  • Applications
    • Real-Time PCR
    • Cloning
    • Protein Biology
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • Septin-8 Antibodies
          Proteintech
          Septin 8 Monoclonal Antibody (4A12G9), CoraLite® Plus 488
          View all (14) Septin-8 antibodies
          Tech Support
          Cite Septin 8 Monoclonal Antibody (4A12G9), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          Septin 8 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Septin 8 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          Septin 8 Antibody (CL488-66188) in ICC/IF
          Immunofluorescent analysis of (-20°C Methanol) fixed HeLa cells using CoraLite® Plus 488 Septin 8 antibody (CL488-66188, Clone: 4A12G9) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Septin 8 Antibody in Immunocytochemistry (ICC/IF)
          Septin 8 Antibody in Immunocytochemistry (ICC/IF)
          Product Details
          CL488-66188
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgM
          Class
          Monoclonal
          Type
          Antibody
          Clone
          4A12G9
          Immunogen
          Septin 8 fusion protein Ag19114
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MKKLDSKVN IIPIIAKADT ISKSELHKFK IKIMGELVSN GVQIYQFPTD DEAVAEINAV MNAHLPFAVV GSTEEVKVGN KLVRARQYPW GVVQVENENH CDFVKLREML IRVNMEDLRE QTHSRHYELY RRCKLEEMGF QDSDGDSQPF SLQETYEAKR KEFLSELQRK EEEMRQMFVN KVKETELELK EKERELHEKF EHLKRVHQEE KRKVEEKRRE LEEETNAFNR RKAAVEALQS QALHATSQQP LRKDKDK
          Target Information
          SEPT8 is a member of the highly conserved septin family. Septins are 40- to 60-kD GTPases that assemble as filamentous scaffolds. They are involved in the organization of submembranous structures, in neuronal polarity, and in vesicle trafficking (Blaser et al., 2003 [PubMed 12909369]).
          Bioinformatics
          Protein Aliases: 2-Sep; CDC10 family member; KIAA0202; septin KIAA0202b; septin KIAA0202c; septin KIAA0202d; Septin-8
          View more View less
          Gene Aliases: KIAA0202; SEP2; SEPT8; Septin-8; SEPTIN8
          View more View less
          UniProt ID: (Human) Q92599
          View more View less
          Entrez Gene ID: (Human) 23176
          View more View less
          Function(s)
          nucleotide binding GTPase activity protein binding GTP binding binding, bridging
          Process(es)
          protein localization regulation of protein stability regulation of intracellular protein transport regulation of SNARE complex assembly cytoskeleton-dependent cytokinesis
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          • Documents and Certificates
          • Study information disclosure
          • Promotions
          • Events and Seminars
          • Blog
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          Japan flag icon
          Japan
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline