  • Applications
    • Real-Time PCR
    • Cloning
    • Protein Biology
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • TLE3 Antibodies
          Proteintech
          TLE3 Recombinant Rabbit Monoclonal Antibody (240680G3), CoraLite® Plus 488
          View all (6) TLE3 antibodies
          Tech Support
          Cite TLE3 Recombinant Rabbit Monoclonal Antibody (240680G3), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          TLE3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          TLE3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          TLE3 Antibody (CL488-83955-4) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed HepG2 cells using CoraLite Plus 488 TLE3 antibody (CL488-83955-4, Clone: 240680G3 ) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          TLE3 Antibody in Immunocytochemistry (ICC/IF)
          TLE3 Recombinant Rabbit Monoclonal Antibody (240680G3), CoraLite® Plus 488
          Product Details
          CL488-83955-4
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          240680G3
          Immunogen
          TLE3 fusion protein Ag1930
          3D Epitope / Immunogen
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1000 µg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Sequence of this protein is as follows: MYPQGRHPAP HQPGQPGFKF TVAESCDRIK DEFQFLQAQY HSLKVEYDKL ANEKTEMQRH YVMYYEMSYG LNIEMHKQTE IAKRLNTILA QIMPFLSQEH QQQVAQAVER AKQVTMTELN AIIGQQQLQA QHLSHATHGP PVQLPPHPSG LQPPGIPPVT GSSSGLLALG ALGSQAHLTV KDEKNHHELD HRERESSANN SVSPSESLRA SEKHRGSADY SMEAKKRKAE EKDSLSRYDS DGDKSDDLVV DVSNEDPATP RVSPAHSPPE NGLDKARSLK KDAPTSPASV ASSSSTPSSK TKDLGHNDKS STPGLKSNTP TPRNDAPTPG TSTTPGLRSM PGKPPGMDPI
          Target Information
          The TLE genes are differentially expressed and encode nuclear proteins, consistent with the presence of sequence motifs associated with nuclear functions. These genes are the human homologues of Drosophila groucho, a member of the Notch signaling pathway which controls a number of different cell-fate choices in invertebrates and vertebrates.
          Bioinformatics
          Protein Aliases: enhancer of split groucho 3; Enhancer of split groucho-like protein 3; ESG-1; ESG-2; ESG-3; ESG-4; ESG3; Tle1; Tle2; tle4; transducin like enhancer of split 3; transducin-like enhancer of split 3 (E(sp1) homolog, Drosophila); transducin-like enhancer of split 3, homolog of Drosophila E(sp1); Transducin-like enhancer protein 3; unnamed protein product
          View more View less
          Gene Aliases: ESG; ESG3; GRG3; HsT18976; KIAA1547; TLE3
          View more View less
          UniProt ID: (Human) Q04726
          View more View less
          Entrez Gene ID: (Human) 7090
          View more View less
          Function(s)
          transcription corepressor activity protein binding
          Process(es)
          regulation of transcription, DNA-templated signal transduction organ morphogenesis Wnt signaling pathway negative regulation of transcription, DNA-templated negative regulation of canonical Wnt signaling pathway negative regulation of cold-induced thermogenesis
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          • Documents and Certificates
          • Study information disclosure
          • Promotions
          • Events and Seminars
          • Blog
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          Japan flag icon
          Japan
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline