  • Applications
    • Real-Time PCR
    • Cloning
    • Protein Biology
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • On-demand videos
    • FAQs
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • ZNF217 Antibodies
          Proteintech
          ZNF217 Recombinant Rabbit Monoclonal Antibody (241217E3), CoraLite® Plus 488
          View all (25) ZNF217 antibodies
          Tech Support
          Cite ZNF217 Recombinant Rabbit Monoclonal Antibody (241217E3), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          ZNF217 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          ZNF217 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          ZNF217 Antibody (CL488-84276) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed HepG2 cells using CoraLite Plus 488 ZNF217 antibody (CL488-84276, Clone: 241217E3 ) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          ZNF217 Antibody in Immunocytochemistry (ICC/IF)
          ZNF217 Recombinant Rabbit Monoclonal Antibody (241217E3), CoraLite® Plus 488
          Product Details
          CL488-84276
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          241217E3
          Immunogen
          ZNF217 fusion protein Ag19453
          3D Epitope / Immunogen
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1000 µg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Sequence of this protein is as follows: IPSITCPFCT FKTFYPEVLM MHQRLEHKYN PDVHKNCRNK SLLRSRRTGC PPALLGKDVP PLSSFCKPKP KSAFPAQSKS LPSAKGKQSP PGPGKAPLTS GIDSSTLAPS NLKSHRPQQN VGVQGAATRQ QQSEMFPKTS VSPAPDKTKR PETKLKPLPV APSQPTLGSS NINGSIDYPA KNDSPWAPPG RDYFCNRSAS NTAAEFGEPL PKRLKSSVVA LDVDQPGANY RRGYDLPKYH MVRGITSLLP QDCVYPSQAL PPKPRFLSSS EVDSPNVLTV QKPYGGSGPL YTCVPAGSPA SSSTLEGKRP VSYQHLSNSM AQKRNYENFI GNAHYRP
          Target Information
          Binds to the promoters of target genes and functions as repressor. Promotes cell proliferation and antagonizes cell death. Promotes phosphorylation of AKT1 at 'Ser-473'.
          Bioinformatics
          Protein Aliases: Kruppel-like; unnamed protein product; Zinc finger protein 217
          View more View less
          Gene Aliases: ZABC1; ZNF217
          View more View less
          UniProt ID: (Human) O75362
          View more View less
          Entrez Gene ID: (Human) 7764
          View more View less
          Function(s)
          transcription regulatory region sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding transcription factor activity, sequence-specific DNA binding protein binding zinc ion binding metal ion binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter negative regulation of transcription, DNA-templated
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          • Documents and Certificates
          • Study information disclosure
          • Promotions
          • Events and Seminars
          • Blog
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          Japan flag icon
          Japan
          Your items have has been added!
          Host server : magellan-search-blue-fcf5f9d6d-9k2pj:80/100.66.79.197:80.
          git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.51.0-2026.05.21-1.0