  • Applications
    • Real-Time PCR
    • Cloning
    • Protein Biology
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • On-demand videos
    • FAQs
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • HEBP1 Antibodies
          Proteintech
          HEBP1 Recombinant Rabbit Monoclonal Antibody (230156B6), CoraLite® Plus 750
          View all (6) HEBP1 antibodies
          Tech Support
          Cite HEBP1 Recombinant Rabbit Monoclonal Antibody (230156B6), CoraLite® Plus 750
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          HEBP1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          HEBP1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          HEBP1 Antibody (CL750-82950) in WB
          Various lysates were subjected to SDS PAGE followed by western blot with CL750-82996-4 (RILP antibody) at dilution of 1:500 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          HEBP1 Antibody in Western Blot (WB)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          HEBP1 Recombinant Rabbit Monoclonal Antibody (230156B6), CoraLite® Plus 750
          Product Details
          CL750-82950
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:2,000
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Expression System
          CHO cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230156B6
          Immunogen
          HEBP1 fusion protein Ag10538
          3D Epitope / Immunogen
          Conjugate
          CoraLite® Plus 750 CoraLite® Plus 750 CoraLite® Plus 750
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          755/780 nm
          Form
          Liquid
          Concentration
          1000 µg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Sequence of this protein is as follows: MLGMIKNSLF GSVETWPWQV LSKGDKEEVA YEERACEGGK FATVEVTDKP VDEALREAMP KVAKYAGGTN DKGIGMGMTV PISFAVFPNE DGSLQKKLKV WFRIPNQFQS DPPAPSDKSV KIEEREGITV YSMQFGGYAK EADYVAQATR LRAALEGTAT YRGDIYFCTG YDPPMKPYGR RNEIWLLKT
          Target Information
          The full-length protein encoded by this gene is an intracellular tetrapyrrole-binding protein. This protein includes a natural chemoattractant peptide of 21 amino acids at the N-terminus, which is a natural ligand for formyl peptide receptor-like receptor 2 (FPRL2) and promotes calcium mobilization and chemotaxis in monocytes and dendritic cells.
          Bioinformatics
          Protein Aliases: p22 HBP; p22HBP; unnamed protein product
          View more View less
          Gene Aliases: HBP; HEBP; p22HBP
          View more View less
          UniProt ID: (Human) Q9NRV9, (Mouse) Q9R257
          View more View less
          Entrez Gene ID: (Human) 50865, (Mouse) 15199, (Rat) 362454
          View more View less
          Function(s)
          heme binding
          Process(es)
          heme metabolic process circadian rhythm
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          • Documents and Certificates
          • Study information disclosure
          • Promotions
          • Events and Seminars
          • Blog
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          Japan flag icon
          Japan
          Your items have has been added!
          Host server : magellan-search-green-dd86746ff-ngdwv:80/100.66.78.131:80.
          git-commit: 04e44c58f20fafe66519586ffe6db5d8a90e52ce
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.53.1-Offline