Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MGKDYYQTL GLARGASDEE IKRAYRRQAL RYHPDKNKEP GAEEKFKEIA EAYDVLSDPR KREIFDRYGE EGLKGSGPSG GSGGGANGTS FSYTFHGDPH AMFAEFFGGR NPFDTFFGQR NGEEGMDIDD PFSGFPMGMG GFTNVNFGRS RSAQEPARKK QDPPVTHDLR VSLEEIYSGC TKKMKISHKR LNPDGKSIRN EDKILTIEVK KGWKEGTKIT FPKEGDQTSN NIPADIVFVL KDKPHNIFKR DGSDVIYPAR ISLREALCGC TVNVPTLDGR TIPVVFKDVI RPGMRRKVPG EGLPLPKTPE KRGDLIIEFE VIFPERIPQT SRTVLEQVLP I (1-340 aa encoded by BC019827)
TCF-9 (transcription factor 9), also known as C2orf3 (chromosome 2 open reading frame 3), GCF (GC-rich sequence DNA-binding factor) or DNABF, is a 781 amino acid nuclear protein that belongs to the GCF family. Widely expressed, GCF binds the GC-rich sequences of beta-Actin, EGFR and calcium-dependent protease (CANP) promoters. GCF contains multiple phosphoserine and phosphothreonine residues, and two GCF isoforms are produced due to alternative splicing events. GCF is considered a candidate for susceptibility to dyslexia (DYX3) as both genes reside in close proximity on human chromosome 2. Chromosome 2 is the second largest human chromosome and consists of 237 million bases, encodes over 1,400 genes and makes up approximately 8% of the human genome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DnaJ (Hsp40) homolog, subfamily B, member 1; dnaJ protein homolog 1; DNAJB1; heat shock 40 kDa protein 1; Heat shock protein; HSP; HSP 40; human DnaJ protein 1; NDAJB1; radial spoke 16 homolog B; unnamed protein product
Gene Aliases: 0610007I11Rik; DjB1; Hdj1; Hsp40; HSPF1; RSPH16B; Sis1
UniProt ID: (Human) P25685, (Mouse) Q9QYJ3
Entrez Gene ID: (Human) 3337, (Mouse) 81489, (Rat) 361384
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support