Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Sequence of this protein is as follows: LPEPGPQQQT FAVPRPPHQP LQMPKMMPEN QYPSEQRFQR QLSEPCHPFP PQPGVPGDNR PSYHRQMSEP IVPAAPPPPQ GFKQEYHDPL YEHGVPGMPG PPAHGFQSPM
ETS variant 5, transcription factor of the ETS family, divergent member of the winged helix-turn-helix superfamily, expressed in brain, placenta and other tissues involved in development. Tissue specificity: Ubiquitous. ERM is expressed in Sertoli cells and spermatogonial stem cells (SSCs). Studies on knockout mice have shown that it is required for SSC self-renewal but not differentiation, thus affected mice exhibit progressive germ cell depletion.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ETS translocation variant 5; ets variant gene 5; ets variant gene 5 (ets-related molecule); ets-related molecule; Ets-related protein ERM
Gene Aliases: 1110005E01Rik; 8430401F14Rik; ERM; ETV5
UniProt ID: (Human) P41161, (Mouse) Q9CXC9
Entrez Gene ID: (Human) 2119, (Rat) 303828, (Mouse) 104156
Molecular Function: winged helix/forkhead transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support