Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • FGR Antibodies

          Abnova

          FGR Polyclonal Antibody, MaxPab™

          View all (18) FGR antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite FGR Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          FGR Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FGR Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          FGR Antibody (H00002268-B01P) in WB

          Western Blot analysis of FGR expression in transfected 293T cell line by FGR MaxPab polyclonal antibody. Lane 1: FGR transfected lysate(58.19 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          FGR Antibody in Western Blot (WB)
          FGR Polyclonal Antibody, MaxPab™

          Product Details

          H00002268-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          FGR (NP_005239.1, 1 a.a. approximately 529 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MGCVFCKKLE PVATAKEDAG LEGDFRSYGA ADHYGPDPTK ARPASSFAHI PNYSNFSSQA INPGFLDSGT IRGVSGIGVT LFIALYDYEA RTEDDLTFTK GEKFHILNNT EGDWWEARSL SSGKTGCIPS NYVAPVDSIQ AEEWYFGKIG RKDAERQLLS PGNPQGAFLI RESETTKGAY SLSIRDWDQT RGDHVKHYKI RKLDMGGYYI TTRVQFNSVQ ELVQHYMEVN DGLCNLLIAP CTIMKPQTLG LAKDAWEISR SSITLERRLG TGCFGDVWLG TWNGSTKVAV KTLKPGTMSP KAFLEEAQVM KLLRHDKLVQ LYAVVSEEPI YIVTEFMCHG SLLDFLKNPE GQDLRLPQLV DMAAQVAEGM AYMERMNYIH RDLRAANILV GERLACKIAD FGLARLIKDD EYNPCQGSKF PIKWTAPEAA LFGRFTIKSD VWSFGILLTE LITKGRIPYP GMNKREVLEQ VEQGYHMPCP PGCPASLYEA MEQTWRLDPE ERPTFEYLQS FLEDYFTSAE PQYQPGDQT

          Target Information

          FGR is a SRC-related kinase present in hematopoietic cells. FGR signals within multiple pathways including within the integrin pathway and downstream of chemokine and cytokine receptors. This protein contains N-terminal sites for myristylation and palmitoylation, a PTK domain, and SH2 and SH3 domains which are involved in mediating protein-protein interactions with phosphotyrosine-containing and proline-rich motifs, respectively. It localizes to plasma membrane ruffles, and functions as a negative regulator of cell migration and adhesion triggered by the beta-2 integrin signal transduction pathway. Infection with Epstein-Barr virus results in the overexpression of this protein.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: c-fgr protooncogene; c-src-2 proto-oncogene; EC 2.7.10.2; feline Gardner-Rasheed sarcoma viral oncogene homolog; FGR protein; FLJ43153; Gardner-Rasheed feline sarcoma viral (v-fgr) oncogene homolog; MGC75096; OTTHUMP00000003603; OTTHUMP00000003604; p55-c-fgr protein; p55-Fgr; p58-Fgr; p58c-Fgr; Proto-oncogene c-Fgr; proto-oncogene tyrosine-protein kinase FGR; Tyrosine-protein kinase Fgr; unnamed protein product; v-fgr feline Gardner-Rasheed sarcoma viral oncogene homolog

          View more View less

          Gene Aliases: c-fgr; c-src2; FGR; p55-Fgr; p55c-fgr; p58-Fgr; p58c-fgr; SRC2

          View more View less

          UniProt ID: (Human) P09769

          View more View less

          Entrez Gene ID: (Human) 2268

          View more View less

          Function(s)
          nucleotide binding phosphotyrosine binding protein kinase activity protein tyrosine kinase activity non-membrane spanning protein tyrosine kinase activity receptor binding protein binding ATP binding kinase activity transferase activity protein kinase binding immunoglobulin receptor binding Fc-gamma receptor I complex binding
          Process(es)
          positive regulation of cytokine production immune system process immune response-regulating cell surface receptor signaling pathway negative regulation of inflammatory response to antigenic stimulus protein phosphorylation transmembrane receptor protein tyrosine kinase signaling pathway integrin-mediated signaling pathway regulation of cell shape response to virus peptidyl-tyrosine phosphorylation cell differentiation bone mineralization positive regulation of cell migration negative regulation of natural killer cell activation Fc-gamma receptor signaling pathway involved in phagocytosis positive regulation of mast cell degranulation innate immune response regulation of innate immune response regulation of protein kinase activity protein autophosphorylation skeletal system morphogenesis regulation of phagocytosis defense response to Gram-positive bacterium positive regulation of protein kinase B signaling
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline