Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • FRMPD1 Antibodies

          Abnova

          FRMPD1 Polyclonal Antibody, MaxPab™

          View all (5) FRMPD1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite FRMPD1 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          FRMPD1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FRMPD1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          FRMPD1 Antibody (H00022844-B01P) in WB

          Western Blot analysis of FRMPD1 expression in transfected 293T cell line (H00022844-T01) by FRMPD1 MaxPab polyclonal antibody.

          Lane 1: FRMPD1 transfected lysate(173.58 KDa).
          Lane 2: Non-transfected lysate.
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          FRMPD1 Antibody in Western Blot (WB)
          FRMPD1 Polyclonal Antibody, MaxPab™

          Product Details

          H00022844-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          FRMPD1 (NP_055722.1, 1 a.a. approximately 1578 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Amount

          50 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MEELETSLFQ TRKAHRIEQM VARWLRRSRD SSARAKVAAA DGPARNPTQT LIPVRHTVKI DKDTLLQDYG FHISESLPLT VVAVTAGGSA HGKLFPGDQI LQMNNEPAED LSWERAVDIL REAEDSLSIT VVRCTSGVPK SSFLTEEKRA RLKTNPVKVH FAEEVLISGH SQGNSLLCMP NVLKLYLENG QTKAFKFEAN TTVKDIILTV KEKLSIRSIE YFALALEEQY SISRLHLLHE EELIQQVVER EESHDYRCLF RVCFVPKDPL DLLKEDPVAF EYLYLQSCSD VLQERFAVEM KCSSALRLAA LHIQERIYAC AQPQKISLKY IEKDWGIENF ISPTLLRNMK GKDIKKAISF HMKRNQNLLE PRQKQLISAA QLRLNYLQIL GELKTYGGRI FNATLMLQDR ESYIALLVGA KYGISQVINS KLNIMSTLAE FANISRVELT EESEKVSVVK VYLQDVKVLT LLLESNSAKD LACLIAGYYR LLVDPVTSIF LWPGNKQQAH RVSAEEGYES RACSDSEESS EVDCVLEPLS DRRLVKLAPC RSLIKEEQPP GNSPTPEVAR RGPSTCGASS TTDSAESEAS DSANTESRGY RTSGSSESMD ALEEDDLDTC SSSRSTFFHF GSPGLAESID SDSQEERSGI ETSGFLCLLD LAQRANPQCQ KTEFSESAAL ETFGWAPELS TVRLDPRLYE GSHADYYSLC SSVSPASYLS DSSESTASRQ GGAPPAWGQQ GWTEAQPSSM LEPLALHPPL AFEDGSSDEE YYDAADKLTP PGPPSGPRDV STAEPSATSL QNKASTSSPE NSLPCGPDGR QPSRRGGVKK YAKTLRKRRS FLQTDYTSQV SFPLVPSASL ESVDDVCYYD REPYLALGAP SPTVSSLQDM QGEPGLLETK ALGLLAPLRE TKSTNPASRV MEMEPETMET KSVIDSRVSS ISAIRFRIDP NNKENSGVVP AASSSASTPH CSNPGSSGPD TAQARPSQIL PLSQDLDGIA PKEPTIEHGD SSFSLSSGDP NPDRACLASN PGLNNVSQGD TLELQLEPHV QLEMGLESFC TNHIQETAPK YTEPLLSPRD EPRSDECGIN PGEKIASIPT KEEPQGQLSL ERDREVTNKN GTNVFQEESR KDSGDSPGDV SNNVSQTLDI SSPAGKIVTS LSLDAPVTGT EQIPPHPPRD PQGQSREPPG QGCQAQEQKL FVELDLDPDF FLGKQTVSPA VPPEGIKAEA PNHVTGQDIA PRDSPEWVCF NPEPSLPEPL PCPQEDPHLE TSNHCLLLEG KSDSSSICLS AEKSFLCFAP ESHPEVSASL RVATSLGFAG MNEMVAPRIG MDQCSCQFSY ATCFRGPQPE TEEEDRDLEA HPMAPLTSPP SAGSPVVLPW RPARAHSCTT APLSRKSHIW PEYCSRALRQ LKATPASTPE GFIQLMESLL ELQDILETSW GVGNKHPPEK CTWHFTESRS RLCMGSQKLL SSCRHVIRMD QSPEEMQGAV RDTFQHLVQL AGLCFQFTDC SRCSARHREA AGNLRDVVYT YHQFIEAAKS TCERGYHDLS VKLLARQCTA LTAAVFCLTQ KFRASTAL

          Target Information

          The FERM and PDZ domain containing (FRMPD) protein family consists of four proteins that contain a FERM (Four-point-one, erzin, radixin, moesin) domain and at least one PDZ (PSD-95/Discs large/Zonula-occuldens-1) domain. FRMPD1 was initially identified through a yeast two-hybrid screening with Activator of G-protein signaling 3 (AGS3), a protein that acts to stabilize the GDP-bound conformation of Gai, as the bait. FRMPD1 binds to the tetratricopeptide repeat of AGS3 and stabilizes AGS3 in a membrane environment. Suppression of FRMPD1 in Cath.a-differentiated neuronal cells decreased the level of endogenous AGS3 in membrane fractions by ~50% and enhanced the inhibition of forskolin-induced increases in cAMP. Co-immunoprecipitation studies indicate that the interaction of AGS3 with FRMPD1 is mutually exclusive with the interaction of AGS3 with Gai3, suggesting that FRMPD1 may position AGS3 in the membrane where it can then interact with Galphai.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: FERM and PDZ domain-containing protein 1; FERM domain-containing protein 2; RP11-263I4.1; unnamed protein product

          View more View less

          Gene Aliases: FRMD2; FRMPD1; KIAA0967

          View more View less

          UniProt ID: (Human) Q5SYB0

          View more View less

          Entrez Gene ID: (Human) 22844

          View more View less

          Function(s)
          protein binding
          Process(es)
          regulation of G-protein coupled receptor protein signaling pathway response to light intensity protein transport light adaption establishment of protein localization establishment of protein localization to membrane
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5btrv:80/100.66.77.21:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline