  • Applications
    • Real-Time PCR
    • Cloning
    • Protein Biology
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • On-demand videos
    • FAQs
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • CD7 Antibodies
          Invitrogen
          CD7 Monoclonal Antibody (MEM-186), PerCP-Cyanine5.5
          View all (104) CD7 antibodies
          Tech Support
          Cite CD7 Monoclonal Antibody (MEM-186), PerCP-Cyanine5.5
          Additional Information:
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          CD7 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD7 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          CD7 Antibody (MA574781) in Flow
          Flow cytometry analysis of CD7 in human peripheral blood cells. Samples were incubated with monoclonal CD7 (Product # MA574781), followed by PerCP-Cy5.5. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CD7 Antibody in Flow Cytometry (Flow)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          CD7 Monoclonal Antibody (MEM-186), PerCP-Cyanine5.5
          Product Details
          MA574781
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          4 µL/10^6 cells
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG1, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          MEM-186
          Immunogen
          Human acute myelogenous leukaemia cell line KG-1.
          3D Epitope / Immunogen
          Conjugate
          PerCP-Cyanine5.5 PerCP-Cyanine5.5 PerCP-Cyanine5.5
          • Unconjugated
          • APC 
          • FITC
          • Pacific Blue
          • PE 
          • PE-DyLight 594
          • PerCP 
          • Request custom conjugation
          Excitation/Emission Max
          489/679 nm View spectra spectra
          Form
          Liquid
          Purification
          Size-exclusion chromatography
          Storage buffer
          PBS, pH 7.4, with 0.2% BSA
          Contains
          15mM sodium azide
          Storage conditions
          4°C, store in dark, DO NOT FREEZE!
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          CD7, also known as gp40 or Leu9, is a 40 kDa receptor and member of the immunoglobulin gene superfamily. It features an N-terminal region (amino acids 1-107) that is highly homologous to Ig kappa light chains, while its carboxyl-terminal region is proline-rich, forming a stalk from which the Ig domain projects. CD7 is prominently expressed on the majority of immature and mature T lymphocytes, as well as T cell leukemias. It is also found on natural killer cells, a small subpopulation of normal B cells, and malignant B cells. CD7 plays a crucial role in modulating immune cell activity. Cross-linking of surface CD7 enhances T cell and NK cell functions, as evidenced by increased calcium flux, expression of adhesion molecules, cytokine secretion, and proliferation. CD7 directly associates with phosphoinositol 3-kinase, and its ligation induces the production of D-3 phosphoinositides and tyrosine phosphorylation. The expression of CD7 is an important marker in leukemia diagnostics, highlighting its significance in both normal immune function and disease states.
          How to use the Panel Builder
          Bioinformatics
          Protein Aliases: CD7 antigen (p41); p41 protein; T-cell leukemia antigen; T-cell surface antigen Leu-9; unnamed protein product
          View more View less
          Gene Aliases: GP40; LEU-9; Tp40; TP41
          View more View less
          UniProt ID: (Human) P09564
          View more View less
          Entrez Gene ID: (Human) 924
          View more View less
          Function(s)
          transmembrane signaling receptor activity protein binding signaling receptor activity
          Process(es)
          adaptive immune response positive regulation of T cell cytokine production immune response transmembrane receptor protein tyrosine kinase signaling pathway T cell activation
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          • Documents and Certificates
          • Study information disclosure
          • Promotions
          • Events and Seminars
          • Blog
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          Japan flag icon
          Japan
          Your items have has been added!
          Host server : magellan-search-blue-74d7b4b4d9-6qggw:80/100.66.76.149:80.
          git-commit: 7aafa54bd143a040505a257d2a3ebda4095a617f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.56.0-Offline