Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MEPQVTLNV TFKNEIQSFL VSDPENTTWA DIEAMVKVSF DLNTIQIKYL DEENEEVSIN SQGEYEEALK MAVKQGNQLQ MQVHEGHHVV DEAPPPVVGA KRLAARAGKK PLAHYSSLVR VLGSDMKTPE DPAVQSFPLV PCDTDQPQDK PPDWFTSYLE TFREQVVNET VEKLEQKLHE KLVLQNPSLG SCPSEVSMPT SEETLFLPEN QFSWHIACNN CQRRIVGVRY QCSLCPSYNI CEDCEAGPYG HDTNHVLLKL RRPVVGSSEP FCHSKYSTPR LPAALEQVRL QKQVDKNFLK AEKQRLRAEK KQRKAEVKEL KKQLKLHRKI HLWNSIHGLQ SPKSPLGRPE SLLQ (1-353 aa encoded by BC009808)
The protein encoded by this gene was originally identified as an ovarian tumor antigen monitored in ovarian cancer. The encoded protein contains a B-box/coiled coil motif, which is present in many genes with transformation potential, but the function of this protein is unknown. This gene is located on a region of chromosome 17q21.1 that is in close proximity to tumor suppressor gene BRCA1. Three alternatively spliced variants encoding the same protein have been identified for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B-box protein; cell migration-inducing gene 19 protein; IAI3B; KIAA0049; membrane component chromosome 17 surface marker 2 homolog; membrane component, chromosome 17, surface marker 2 (ovarian carcinoma antigen CA125); MIG19; migration-inducing protein 19; neighbor of BRCA1 gene 1; neighbor of BRCA1 gene 1 protein; next to the Brca1; ovarian carcinoma antigen CA125; unnamed protein product
Gene Aliases: 1A1-3B; Ca125; IAI3B; M17S2; MIG19; mKIAA0049; RGD1311421
UniProt ID: (Human) Q14596, (Mouse) P97432, (Rat) Q501R9
Entrez Gene ID: (Human) 4077, (Mouse) 17966, (Rat) 303554
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support