Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • SCN5A Antibodies

          Alomone Labs, LTD

          NaV1.5 (SCN5A) Polyclonal Antibody

          View all (9) SCN5A antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite NaV1.5 (SCN5A) Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          NaV1.5 (SCN5A) Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.
          NaV1.5 (SCN5A) Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NaV1.5 (SCN5A) Antibody (ASC-005-GP200UL) in IHC (F)

          Expression of NaV1.5 in rat heart - Immunohistochemical staining of rat heart frozen sections using Guinea pig Anti-NaV1.5 (SCN5A) Antibody (#ASC-005-GP). NaV1.5 staining (red) appears in cardiomyocytes and in intercalated discs (arrows). Nuclei are stained using DAPI (blue). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NaV1.5 (SCN5A) Antibody in Immunohistochemistry (Frozen) (IHC (F))
          NaV1.5 (SCN5A) Antibody in Western Blot (WB)
          NaV1.5 (SCN5A) Polyclonal Antibody

          Product Details

          ASC-005-GP200UL

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:200
          -

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Guinea pig / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          DRLPKSDSEDGPRALNQLSC, corresponding to amino acid residues 493-511 of rat NaV1.5, Intracellular loop between domains I and II
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          0.95 mg/mL

          Amount

          190 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 1% BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).

          Target Information

          Scn5a is an integral membrane protein and tetrodotoxin-resistant voltage-gated sodium channel subunit. Scn5a is found primarily in cardiac muscle and is responsible for the initial upstroke of the action potential in an electrocardiogram. Defects in Scn5a are a cause of long QT syndrome type 3 (LQT3), an autosomal dominant cardiac disease. Alternative splicing results in several transcript variants encoding different isoforms of Scn5a. Voltage-gated sodium channesl (VGSCs) mediate regenerative cell membrane depolarization and conduction of electrical signalling in nerves and muscles. Expression of Scn5a is also detected in lymphocytes, glia, and fibroblasts. Further, Scn5a mediates the voltage-dependent sodium ion permeability of excitable membranes. Assuming opened or closed conformations in response to the voltage difference across the membrane, Scn5a forms a sodium-selective channel through which Na(+) ions may pass in accordance with their electrochemical gradient. Functionally, Scn5a is responsible for the initial upstroke of the action potential and the channel inactivation is regulated by intracellular calcium levels. Voltage-gated sodium channels (NaV) are responsible for action potential initiation and propagation in excitable cells, including nerve, muscle, and neuroendocrine cell types. Scn5a are also expressed at low levels in non-excitable cells, where their physiological role is unclear. Diseases associated with SCN5A include Long Qt Syndrome-3 and Brugada Syndrome 1.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: cardiac tetrodotoxin-insensitive voltage-dependent sodium channel alpha subunit; CDCD2VF1; CIN5; H1B; hH1; hH1c; voltage-gated sodium channel; HH1CMD1E; LQT3SSS1CMPD2; mH1; Nav1.5/SCN5A.3; RATRSKM2X; RH1; RSkM2; RSKM2X; Scn2x; scn5a-JEJ, HH1, hNav1.5, LQT3; Sodium channel protein cardiac muscle subunit alpha; Sodium channel protein type 5 subunit alpha; Sodium channel protein type V subunit alpha; sodium channel voltage-gated type V alpha polypeptide; sodium channel, voltage-gated, type 5, alpha subunit; sodium channel, voltage-gated, type V, alpha polypeptide; sodium channel, voltage-gated, type V, alpha subunit; voltage-gated sodium channel Nav1.5c; Voltage-gated sodium channel subunit alpha Nav1.5

          View more View less

          Gene Aliases: CDCD2; CMD1E; CMPD2; HB1; HB2; HBBD; HH1; ICCD; IVF; LQT3; mH1; Nav1.5; Nav1.5c; PFHB1; SCAL; SCN5A; SkM1; SkM2; SSS1; VF1

          View more View less

          UniProt ID: (Human) Q14524, (Rat) P15389, (Mouse) Q9JJV9

          View more View less

          Entrez Gene ID: (Human) 6331, (Rat) 25665, (Mouse) 20271

          View more View less

          Function(s)
          ion channel activity voltage-gated sodium channel activity cation channel activity sodium channel activity protein binding calmodulin binding fibroblast growth factor binding enzyme binding protein kinase binding protein domain specific binding ankyrin binding ubiquitin protein ligase binding ion channel binding nitric-oxide synthase binding voltage-gated sodium channel activity involved in cardiac muscle cell action potential voltage-gated sodium channel activity involved in AV node cell action potential voltage-gated sodium channel activity involved in bundle of His cell action potential voltage-gated sodium channel activity involved in Purkinje myocyte action potential voltage-gated sodium channel activity involved in SA node cell action potential scaffold protein binding voltage-gated ion channel activity voltage-gated ion channel
          Process(es)
          regulation of heart rate cardiac conduction system development cardiac ventricle development brainstem development ion transport sodium ion transport positive regulation of sodium ion transport response to denervation involved in regulation of muscle adaptation telencephalon development cerebellum development ion transmembrane transport sodium ion transmembrane transport odontogenesis of dentin-containing tooth positive regulation of action potential positive regulation of epithelial cell proliferation membrane depolarization transmembrane transport regulation of cardiac muscle contraction cardiac muscle contraction regulation of ventricular cardiac muscle cell membrane repolarization regulation of atrial cardiac muscle cell membrane depolarization regulation of atrial cardiac muscle cell membrane repolarization regulation of ventricular cardiac muscle cell membrane depolarization cellular response to calcium ion cardiac muscle cell action potential involved in contraction regulation of cardiac muscle cell contraction ventricular cardiac muscle cell action potential membrane depolarization during action potential membrane depolarization during cardiac muscle cell action potential atrial cardiac muscle cell action potential SA node cell action potential AV node cell action potential bundle of His cell action potential membrane depolarization during AV node cell action potential membrane depolarization during SA node cell action potential membrane depolarization during Purkinje myocyte cell action potential membrane depolarization during bundle of His cell action potential AV node cell to bundle of His cell communication regulation of heart rate by cardiac conduction membrane depolarization during atrial cardiac muscle cell action potential regulation of sodium ion transmembrane transport positive regulation of heart rate response to organic cyclic compound neuronal action potential regulation of ion transmembrane transport bundle of His cell to Purkinje myocyte communication SA node cell to atrial cardiac muscle cell communication transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline