  • Applications
    • Real-Time PCR
    • Cloning
    • Protein Biology
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • Defensin alpha-3 Antibodies
          Invitrogen
          Defensin alpha-3 Polyclonal Antibody
          View all (4) Defensin alpha-3 antibodies
          Tech Support
          Cite Defensin alpha-3 Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (10)
          Defensin alpha-3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Defensin alpha-3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FIGURE: 1 / 10
          Defensin alpha-3 Antibody (PA5-122038) in IHC (P)
          Immunohistochemistry (Paraffin) analysis of Defensin alpha-3 in human liver cancer tissue using Defensin alpha-3 Polyclonal Antibody (Product # PA5-119080) (30 µg/mL) and using PBS intead of the primary antibody as a control. The sample was then stained with HRP-Goat anti-Rabbit IgG polyclonal secondary antibody (2 µg/mL). DAB staining. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Defensin alpha-3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Defensin alpha-3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Defensin alpha-3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Defensin alpha-3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Defensin alpha-3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Defensin alpha-3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Defensin alpha-3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Defensin alpha-3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Defensin alpha-3 Antibody in Western Blot (WB)
          Defensin alpha-3 Antibody in Western Blot (WB)
          Defensin alpha-3 Polyclonal Antibody
          Product Details
          PA5-122038
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          0.01-3 µg/mL
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          5-30 µg/mL
          -
          Product Specifications
          Species Reactivity
          Human, Pig
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Recombinant protein Defensin Alpha 3, Neutrophil Specific. The antigen corresponds to amino acid range 21-94 of the target protein.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          10 µg
          Purification
          Antigen affinity chromatography, Protein A
          Storage buffer
          PBS, pH 7.4, with 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2915610
          Target Information
          Defensin alpha-1 is part of a family of antimicrobial and cytotoxic peptides thought to be involved in host defense. Defensins are abundant in the granules of neutrophils and also found in the epithelia of mucosal surfaces such as those of the intestine, respiratory tract, urinary tract, and vagina. Members of the defensin family are highly similar in protein sequence and distinguished by a conserved cysteine motif. The Defensin alpha-1 protein is encoded by the DFA1 gene and is found in the microbicidal granules of neutrophils and likely plays a role in phagocyte-mediated host defense. Several alpha defensin genes are clustered on chromosome 8, and this gene differs from defensin, alpha 3 by only one amino acid. The DFA1 gene and the gene encoding defensin, alpha 3 are both subject to copy number variation. Two transcript variants encoding different isoforms of Defensin alpha-1 have been found.
          Bioinformatics
          Protein Aliases: defensin 3; defensin 3, neutrophil-specific; defensin precursor; Defensin, alpha 3; defensin, alpha 3, neutrophil-specific; HNP-3; HP-3; Neutrophil defensin 3; neutrophil peptide 3; unnamed protein product
          View more View less
          Gene Aliases: DEF3; DEFA3; HNP-3; HNP3; HP-3; HP3
          View more View less
          UniProt ID: (Human) P59666
          View more View less
          Entrez Gene ID: (Human) 1668
          View more View less
          Function(s)
          protein homodimerization activity pore-forming activity
          Process(es)
          innate immune response in mucosa defense response antibacterial humoral response intracellular estrogen receptor signaling pathway killing of cells of other organism defense response to bacterium innate immune response defense response to Gram-negative bacterium defense response to Gram-positive bacterium defense response to fungus defense response to virus membrane disruption in other organism antimicrobial humoral immune response mediated by antimicrobial peptide
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          • Documents and Certificates
          • Study information disclosure
          • Promotions
          • Events and Seminars
          • Blog
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          Japan flag icon
          Japan
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline