Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PDIA6 Antibodies

          Proteintech

          PDIA6 Polyclonal Antibody

          19 References
          View all (19) PDIA6 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite PDIA6 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (22)
          PDIA6 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          PDIA6 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 22

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PDIA6 Antibody (18233-1-AP) in ICC/IF

          Immunofluorescent analysis of HeLa cells using 18233-1-AP (PDIA6 antibody) at dilution of 1:50 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IgG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PDIA6 Antibody in Immunocytochemistry (ICC/IF)
          PDIA6 Antibody in Immunocytochemistry (ICC/IF)
          PDIA6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PDIA6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PDIA6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PDIA6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PDIA6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PDIA6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PDIA6 Antibody in Western Blot (WB)
          PDIA6 Antibody in Western Blot (WB)
          PDIA6 Antibody in Western Blot (WB)
          PDIA6 Antibody in Western Blot (WB)
          PDIA6 Antibody in Western Blot (WB)
          PDIA6 Antibody in Western Blot (WB)
          PDIA6 Antibody in Western Blot (WB)
          PDIA6 Antibody in Western Blot (WB)
          PDIA6 Antibody in Western Blot (WB)
          PDIA6 Antibody in Western Blot (WB)
          PDIA6 Antibody in Western Blot (WB)
          PDIA6 Antibody in Western Blot (WB)
          PDIA6 Antibody in Western Blot (WB)
          PDIA6 Antibody in Immunoprecipitation (IP)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          PDIA6 Polyclonal Antibody

          Product Details

          18233-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:4,000
          View 14 publications 14 publications

          Immunohistochemistry (IHC)

          -
          View 2 publications 2 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:20-1:200
          View 2 publications 2 publications

          Immunoprecipitation (IP)

          0.5-4.0 µg
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          PDIA6 Fusion Protein Ag12940 (1-440 aa encoded by BC001312)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.23 mg/mL

          Amount

          34.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          This antibody is specific to PDIA6.

          Immunogen sequence: MALLVLGLV SCTFFLAVNG LYSSSDDVIE LTPSNFNREV IQSDSLWLVE FYAPWCGHCQ RLTPEWKKAA TALKDVVKVG AVDADKHHSL GGQYGVQGFP TIKIFGSNKN RPEDYQGGRT GEAIVDAALS ALRQLVKDRL GGRSGGYSSG KQGRSDSSSK KDVIELTDDS FDKNVLDSED VWMVEFYAPW CGHCKNLEPE WAAAASEVKE QTKGKVKLAA VDATVNQVLA SRYGIRGFPT IKIFQKGESP VDYDGGRTRS DIVSRALDLF SDNAPPPELL EIINEDIAKR TCEEHQLCVV AVLPHILDTG AAGRNSYLEV LLKLADKYKK KMWGWLWTEA GAQSELETAL GIGGFGYPAM AAINARKMKF ALLKGSFSEQ GINEFLRELS FGRGSTAPVG GGAFPTIVER EPWDGRDGEL PVEDDIDLSD VELDDLGKDE L (1-440 aa encoded by BC001312)

          Target Information

          P5 is a novel protein that was originally identified in humans. Functionally, P5 has been shown to have peptide binding ability and chaperone activity specific to certain proteins. P5 shares several structural similarities to PDI, an important protein of the ER. The deduced amino acid sequence of P5 shows that it contains the ER retention signal KDEL. P5 also contains two CXXC-like motifs, a motif responsible for oxidoreductase activity. These two CXXC motifs are found on the N- and C-terminus of the protein, one at each end. Studies suggest that the N-terminal CXXC motif is more important than the second for isomerase activity. Contrary to most of the PDI-like proteins, P5 is not stress-inducible.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: CaBP1; calcium-binding protein 1; endoplasmic reticulum protein 5; epididymis secretory sperm binding protein; ER protein 5; protein disulfide isomerase associated 6; protein disulfide isomerase P5; protein disulfide isomerase-associated 6; protein disulfide isomerase-related protein; thioredoxin domain containing 7; thioredoxin domain containing 7 (protein disulfide isomerase); thioredoxin domain-containing protein 7; unnamed protein product

          View more View less

          Gene Aliases: 1700015E05Rik; CaBP1; CaBP5; ERP5; P5; TXNDC7

          View more View less

          UniProt ID: (Human) Q15084, (Mouse) Q922R8, (Rat) Q63081

          View more View less

          Entrez Gene ID: (Human) 10130, (Mouse) 71853, (Rat) 286906

          View more View less

          Function(s)
          protein disulfide isomerase activity protein binding protein disulfide oxidoreductase activity calcium ion binding
          Process(es)
          protein folding response to endoplasmic reticulum stress platelet activation IRE1-mediated unfolded protein response platelet aggregation negative regulation of IRE1-mediated unfolded protein response
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-74676f44b6-pqb5n:80/100.66.76.64:80.
          git-commit: db542085ed6beb0c3f0dd774c2548ab49b9f1b6a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026.10.42-1.0