Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • On-demand videos
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • RAPGEF4 Antibodies

          Abnova

          RAPGEF4 Polyclonal Antibody, MaxPab™

          View all (8) RAPGEF4 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite RAPGEF4 Polyclonal Antibody, MaxPab™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          RAPGEF4 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          RAPGEF4 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RAPGEF4 Antibody (H00011069-B01P) in WB

          Western Blot analysis of RAPGEF4 expression in transfected 293T cell line by RAPGEF4 MaxPab polyclonal antibody. Lane 1: RAPGEF4 transfected lysate(111.21 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RAPGEF4 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          H00011069-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          RAPGEF4 (AAH24004.1, 1 a.a. approximately 1011 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MVAAHAAHSS SSAEWIACLD KRPLERSSED VDIIFTRLKE VKAFEKFHPN LLHQICLCGY YENLEKGITL FRQGDIGTNW YAVLAGSLDV KVSETSSHQD AVTICTLGIG TAFGESILDN TPRHATIVTR ESSELLRIEQ KDFKALWEKY RQYMAGLLAP PYGVMETGSN NDRIPDKENT PLIEPHVPLR PANTITKVPS EKILRAGKIL RNAILSRAPH MIRDRKYHLK TYRQCCVGTE LVDWMMQQTP CVHSRTQAVG MWQVLLEDGV LNHVDQEHHF QDKYLFYRFL DDEHEDAPLP TEEEKKECDE ELQDTMLLLS QMGPDAHMRM ILRKPPGQRT VDDLEIIYEE LLHIKALSHL STTVKRELAG VLIFESHAKG GTVLFNQGEE GTSWYIILKG SVNVVIYGKG VVCTLHEGDD FGKLALVNDA PRAASIVLRE DNCHFLRVDK EDFNRILRDV EANTVRLKEH DQDVLVLEKV PAGNRASNQG NSQPQQKYTV MSGTPEKILE HFLETIRLEA TLNEATDSVL NDFIMMHCVF MPNTQLCPAL VAHYHAQPSQ GTEQEKMDYA LNNKRRVIRL VLQWAAMYGD LLQEDDVSMA FLEEFYVSVS DDARMIAALK EQLPELEKIV KQISEDAKAP QKKHKVLLQQ FNTGDERAQK RQPIRGSDEV LFKVYCMDPT YTTIRVPVAT SVKEVISAVA DKLGSGEGLI IVKMSSGGEK VVLKPNDVSV FMTLTINGRL FACPREQFDS LTPLPEQEGP TVGTVGTFEL MSSKDLAYQM TIYDWELFNC VHELELIYHT FGRHNFKKTT ANLDLFLRRF NEIQFWVVTE ICLCSQLSKR VQLLKKFIKI AAHCKEYKNL NSFFAIVMGL SNVAVSRLAL TWEKLPSKFK KFYAEFESLM DPSRNHRAYR LTVAKLEPPL IPFMPLLIKD MTFTHEGNKT FIDNLVNFEK MRMIANTART VRYYRSQPFN PDAAQANKNH QDVRSYVRQL NVIDNQRTLS QMSHRLEPRR P

          Target Information

          Guanine nucleotide exchange factor (GEF) for RAP1A, RAP1B and RAP2A small GTPases that is activated by binding cAMP. Seems not to activate RAB3A. Involved in cAMP-dependent, PKA-independent exocytosis through interaction with RIMS2.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: cAMP-regulated guanine nucleotide exchange factor II; exchange factor directly activated by cAMP 2; exchange protein directly activated by cAMP 2; gefii; putative protein product of Nbla00496; Rap guanine nucleotide exchange factor (GEF) 4; RAP guanine-nucleotide-exchange factor (GEF) 4; unnamed protein product

          View more View less

          Gene Aliases: CAMP-GEFII; CGEF2; EPAC; EPAC 2; EPAC2; Nbla00496

          View more View less

          UniProt ID: (Human) Q8WZA2

          View more View less

          Entrez Gene ID: (Human) 11069

          View more View less

          Function(s)
          guanyl-nucleotide exchange factor activity protein binding cAMP binding protein binding, bridging small GTPase binding
          Process(es)
          adaptive immune response G-protein coupled receptor signaling pathway small GTPase mediated signal transduction positive regulation of insulin secretion Rap protein signal transduction intracellular signal transduction regulation of synaptic vesicle cycle
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-8cpz7:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch