Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • RORA Antibodies

          Bioss

          ROR alpha Polyclonal Antibody

          View all (16) RORA antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite ROR alpha Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          ROR alpha Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          ROR alpha Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ROR alpha Antibody (BS-20687R) in WB

          Mouse kidney lysates probed with ROR alpha Polyclonal Antibody, Unconjugated (bs-20687R) at 1:1000 dilution and 4°C overnight incubation. Followed by conjugated secondary antibody incubation at 1:20000 for 60 min at 37°C. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ROR alpha Antibody in Western Blot (WB)
          ROR alpha Antibody in Western Blot (WB)
          ROR alpha Antibody in Western Blot (WB)
          ROR alpha Polyclonal Antibody

          Product Details

          BS-20687R

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          ELISA (ELISA)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          KLH conjugated synthetic peptide derived from human ROR alpha, amino acids 321-420.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          0.01M TBS, pH 7.4, with 50% glycerol, 1% BSA

          Contains

          0.02% ProClin 300

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          RORA is a receptor for retinoic acid belonging to the NR1 subfamily of nuclear hormone receptors with a nuclear receptor DNA binding domain. RORA binds either as a monomer or as a homodimer to the retinoic acid response element and thus regulates gene expression and also controls cell function. Though the exact function of RORA is yet unknown, it interacts with NM23-2, a nucleoside diphosphate kinase involved in organogenesis and differentiation, NM23-1, the product of a tumor metastasis suppressor candidate gene, CDK7, NCOA3 and NCOA6 coactivators. It is found to express in mesenchymal stem cells derived from bone marrow and are known to act by direct modulation of a bone matrix component. It is widely expressed in most of the tissues. Chromosomal aberrations involving RORA may be a cause of acute promyelocytic leukemia (APL).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Nuclear receptor ROR-alpha; Nuclear receptor RZR-alpha; Nuclear receptor subfamily 1 group F member 1; putative new variant; RAR-related orphan nuclear receptor alpha; RAR-related orphan receptor A; RAR-related orphan receptor alpha; retinoic acid receptor-related orphan receptor alpha; retinoid-related orphan receptor alpha; Retinoid-related orphan receptor-alpha; ROR Alpha; ROR-alpha; Roralpha; transcription factor RZR-alpha

          View more View less

          Gene Aliases: 9530021D13Rik; IDDECA; nmf267; NR1F1; ROR1; ROR2; ROR3; RORA; RORa1; RORalpha; RZR-ALPHA; RZRA; sg; staggerer; tmgc26

          View more View less

          UniProt ID: (Human) P35398, (Mouse) P51448

          View more View less

          Entrez Gene ID: (Human) 6095, (Rat) 300807, (Mouse) 19883

          View more View less

          Function(s)
          RNA polymerase II regulatory region sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcription cofactor binding transcription corepressor binding transcription coactivator binding DNA binding transcription factor activity, sequence-specific DNA binding RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding protein binding beta-catenin binding oxysterol binding zinc ion binding sequence-specific DNA binding metal ion binding transcription factor activity, direct ligand regulated sequence-specific DNA binding core promoter sequence-specific DNA binding steroid hormone receptor activity transcription factor binding C4 zinc finger nuclear receptor
          Process(es)
          angiogenesis regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter xenobiotic metabolic process nitric oxide biosynthetic process circadian rhythm regulation of smoothened signaling pathway regulation of gene expression positive regulation of vascular endothelial growth factor production regulation of glucose metabolic process regulation of steroid metabolic process cerebellar Purkinje cell differentiation cerebellar granule cell precursor proliferation intracellular receptor signaling pathway circadian regulation of gene expression cellular response to sterol cholesterol homeostasis muscle cell differentiation regulation of circadian rhythm positive regulation of circadian rhythm regulation of macrophage activation negative regulation of I-kappaB kinase/NF-kappaB signaling negative regulation of fat cell differentiation positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter cGMP metabolic process rhythmic process negative regulation of inflammatory response triglyceride homeostasis cellular response to interleukin-1 cellular response to tumor necrosis factor cellular response to hypoxia T-helper 17 cell differentiation transcription, DNA-templated steroid hormone mediated signaling pathway regulation of transcription involved in cell fate commitment regulation of cholesterol homeostasis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline