Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
The calculated molecular weight of ZEB1 is 124 kDa, but the post phosphorylation of protein is about 190-210 kDa. This is a rabbit polyclonal antibody raised against part of the full length of ZEB1 of human.
Immunogen sequence: MVQAVVLPT VGLVSPISIN LSDIQNVLKV AVDGNVIRQV LENNQANLAS KEQETINASP IQQGGHSVIS AISLPLVDQD GTTKIIINYS LEQPSQLQVV PQNLKKENPV ATNSCKSEKL PEDLTVKSEK DKSFEGGVND STCLLCDDCP GDINALPELK HYDLKQPTQP PPLPAAEAEK PESSVSSATG DGNLSPSQPP LKNLLSLLKA YYALNAQPSA EELSKIADSV NLPLDVVKKW FEKMQAGQIS VQSSEPSSPE PGKVNIPAKN NDQPQSANAN EPQDSTVNLQ SPLKMTNSPV LPVGSTTNGS RSSTPSPSPL NLSSSRNTQG YLYTAEGAQE EPQVEPLDLS L (391-740 aa encoded by BC112392)
ZEB1 (Zinc finger E-box-binding homeobox 1) acts as a transcriptional repressor. It inhibits interleukin-2 (IL-2) gene expression. ZEB1 enhances or represses the promoter activity of the ATP1A1 gene depending on the quantity of cDNA and depending on the cell type. It represses E-cadherin promoters and induces an epithelial-mesenchymal transition (EMT) by recruiting SMARCA4/BRG1. ZEB1 also represses BCL6 transcription in the presence of the corepressor CTBP1. It postively regulates neuronal differentiation and represses RCOR1 transcription activation during neurogenesis. Mutations in the gene can result in corneal dystrophy, posterior polymorphous and corneal dystrophy, Fuchs endothelial 6.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: delta-crystallin enhancer binding factor 1; DELTA-EF1; MGC133261; negative regulator of IL2; NIL-2-A; NIL-2A; OTTHUMP00000019405; posterior polymorphous corneal dystrophy 3; RP11-472N13.4; transcription factor 8; transcription factor 8 (represses interleukin 2 expression); twirler; unnamed protein product; zinc finger homeobox protein 1a; zinc finger homeodomain enhancer-binding protein
Gene Aliases: 3110032K11Rik; [delta]EF1; AREB6; BZP; DELTAEF1; FECD6; MEB1; Nil2; NIL2A; PPCD3; TCF-8; Tcf18; TCF8; Tw; ZEB; ZFHEP; ZFHX1A; Zfx1a; Zfx1ha
UniProt ID: (Human) P37275, (Mouse) Q64318
Entrez Gene ID: (Human) 6935, (Mouse) 21417
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support