Search
![]() Countess 3自動細胞計數儀 |
![]() Countess 3 FL自動細胞計數儀 |
| 產品概要 | |
|---|---|
| 高準確度、易於使用的明視野細胞計數儀,且兼容於可重複使用玻片 |
高準確度、易於使用的明視野和螢光細胞計數儀,配備可更換的光源模組,且兼容於可重複使用玻片
|
| 應用範例 | |
| 細胞培養室,需求為準確、簡便、快速且經濟實惠的明視野細胞計數,以便為下游應用和細胞繼代培養做準備。 |
細胞生物學實驗室,需求為準確、簡便、快速且經濟實惠的明視野和螢光細胞計數,以擴展到對混合細胞群進行深入分析,例如區分活細胞與死細胞、有核與無核、轉染與未轉染等。
|
While the Countess II/II FL models are no longer available, the Countess 3/3 FL boasts a myriad of enhanced features designed to help streamline cell counting processes with exceptional accuracy and precision. Explore the detailed comparison between Countess II/II FL and Countess 3/3 FL Automated cell counters below.
| 特點 | Countess 3自動細胞計數儀 |
Countess 3 FL自動細胞計數儀 |
機器學習演算法 |
|
|
| 1280 x 800像素 LCD液晶螢幕 | ||
| Wi-Fi雲端連接 | ||
明視野影像和計數 |
||
| 明視野自動亮度調整 | ||
| 明視野快速影像擷取 | ||
| 螢光影像和計數 | 不適用 | 兩個可更換的螢光通道 (多種光源模組供用戶選擇) |
| 螢光自動亮度調整 | 不適用 | |
| 過曝像素指示 | 不適用 | |
| 自動對焦 | ||
自動儲存 |
||
| Countess細胞計數Chamber slides | ||
Countess可重複使用玻片 |
||
預稀釋計算器 |
||
| 細胞繼代計算器 | ||
| 單細胞數據 | ||
21 CFR Part 11合規性的支持軟體 |
||
| FCS格式文件 |
* Countess 3自動細胞計數儀的新特色
Both Countess 3 models work the same way and share common features:
Both models accurately count “difficult” samples such as those containing clumpy cells or debris, and both reduce variability between users, helping increase reproducibility.
The key difference between the models is that the Countess 3 instrument counts in brightfield mode only, while the Countess 3 FL instrument can count in either brightfield only or brightfield and fluorescence mode using interchangeable EVOS LED light cubes. Fluorescence capabilities open up additional application options for the Countess 3 FL instrument, and can extend your research beyond just GFP and RFP with 14 EVOS LED light cubes to choose from.
Looking for information about Countess II Automated Cell Counters? See Countess II resources.
僅供研究用途,不得用於診斷程序。