Accelerate Analysis of Trace Elements for Routine Laboratory
icap-pro-xps-front-rightThe days of limiting your routine trace elemental analysis workflow productivity are over. The launch of a new series of inductively coupled plasma optical emission spectroscopy (ICP-OES) instruments for food and beverage, consumer safety, industrial, environmental and pharmaceutical laboratories is streamlining workflows.
The Thermo Scientific iCAP PRO Series ICP-OES platform is designed to provide a fast, sensitive range of trace element analysis solutions capable of capturing the complete spectrum of high matrix samples in a single run, improving workflow productivity and reducing analysis costs. From standby to start-up in just five minutes, it reduces gas consumption within a vertical dual-purged optical pathway interface that has lower installation requirements due to its standard wall socket and low extraction flow rate.
By using the iCAP PRO Series ICP-OES instruments you can:
    • Reduce the number of measurements per run to obtain a complete spectrum through the combination of a single optical slit and a charge injection device detector, delivering increased analysis speeds and readout in a reduced time frame.
    • Maximize instrument uptime with fast start-up, making analysis possible after just five minutes.
    • Experience minimal recalibration of the instrument as a result of highly stable optics and polychromator.
Learn more about the iCAP PRO Series ICP-OES platform, here.
AnalyteGuru Staff