Here, we look at why full scan high-resolution accurate mass spectrometry (HRAM) techniques based on Thermo Scientific Orbitrap mass analyzer technologies are proving so effective when it comes to detecting the unexpected.
Full scan HRAM: The benefits of Orbitrap mass analyzers over Q-TOF systems
Full scan (high-resolution accurate mass) HRAM is one of the most efficient and widely used mass spectrometry techniques for characterizing unknown compounds in complex samples. Unlike selected ion monitoring (SIM) approaches, which acquire data for pre-defined target analytes with specific mass-to-charge (m/z) ratios, full scan HRAM approaches enable the collection of data over a much broader m/z range. As a result, all ions and fragments within that mass range (including analytes that aren’t on your target list) can be studied.
The most widely-used instruments for full scan analysis are those based on quadrupole-time-of-flight (Q-TOF) and Orbitrap mass analyzer designs. While Q-TOF mass spectrometers have historically served the analytical community well, remarkable advances in the mass accuracy, resolution and sensitivity of Orbitrap mass analyzer systems over the past decade mean these systems now offer a number of key advantages over Q-TOF systems.
Today, Orbitrap mass analyzer systems can achieve resolutions of up to 480,000 (FWHM) at m/z 200 (with the option of increasing this to up to 1,000,000 FWHM), far beyond the capabilities of Q-TOF systems (Figure 1). This exceptional mass resolution and sub-1 ppm mass accuracy allows analysts to discriminate between analytes of interest and interfering ions over a wide mass range. Distinguishing between ions of interest and interfering ions in the low and very-low m/z range is particularly critical for the confident characterization of a wide range of analytes, including small molecules, metabolites, lipids and peptides as well as their fragment ions.
Another feature of Orbitrap mass analyzer systems is that mass resolution can be easily adjusted, from those typically used for MS2 scans to the ultra-high levels (240,000 FWHM at m/z 200) employed in full scan MS. This flexibility allows analysts to adjust the number of data points across liquid chromatography peaks without losing sensitivity – something that isn’t always the case with Q-TOF systems. Orbitrap mass analyzer instruments such as the Thermo Scientific Q Exactive series of mass spectrometers also guarantee a high linearity range that is independent of resolution and do not suffer from detector saturation effects. This is in contrast to Q-TOF instruments, where rapid saturation of the detector can cause issues with linearity of response.
Why is high mass resolution so important when characterizing unknown analytes?
High mass resolution is critical for isotopic distribution assignment. Fine isotope assignment is a powerful tool for analyte confirmation as this information gives additional confidence in the identification of unknowns and can even be used to partially replace the information derived from MS2 fragments when their intensity is insufficient to be detected or when precursor ions have poor fragmentation.
Here, high mass accuracy and mass resolution are useful for two reasons: the accurate determination of the monoisotopic mass peak restricts the number of possible elemental compositions, while high resolution in combination with accurate mass measurements allow fine structures to be revealed, further narrowing the range of identities. When combined with Thermo Scientific Compound Discoverer or Thermo Fisher Mass Frontier spectra interpretation data analysis software and spectral database searching using mzCloud mass spectral library for fragment ions and/or ChemSpider for intact compounds, high resolution, high mass accuracy data can reveal the identity of unknown compounds.
Characterizing unknown analytes can be one of the most challenging applications in analytical laboratory workflows. However, thanks to the high and ultra-high mass resolution capabilities of Orbitrap instruments, non-targeted analysis is becoming as easy and convenient as targeted screening.
Released in 2018, the Thermo Scientific Orbitrap ID-X Tribrid Mass Spectrometer combines quadrupole, Orbitrap and linear ion trap mass analyzer technology, enabling users to acquire data with less experimental setup and interpretation expertise. Scientists can now leverage an intelligent data acquisition and analysis strategy to enhance their interpretation of an unknown compound’s mass spectra, helping relieve laboratory bottlenecks that limit the characterization, elucidation and identification of small molecules across extractables and leachables, metabolite ID, metabolomics, lipidomics and food contact material applications.
Want to learn more about Orbitrap mass analyzers?
Find out more about the power of HRAM using Orbitrap mass analyzer technology using the resources below:
-
- Compendium: Orbitrap Quantitation: Lab of the Future
-
- Whitepaper: Compounding insights for small molecule research