Responding to Hantavirus: Advancing Rapid Molecular Detection
Article Summary
Laboratories responding to the emerging 2026 Andes hantavirus incident need rapid, scalable molecular workflows that can integrate into existing RT-qPCR infrastructure. This workflow is best suited for public health laboratories, surveillance programs, academic research labs, and high-throughput testing facilities seeking rapid research-use-only (RUO) molecular detection capabilities. By combining optimized RT-qPCR assays, extraction workflows, and broad instrument compatibility, laboratories can strengthen preparedness for emerging infectious disease research.
Why the 2026 Andes Virus Incident Matters
Recent developments in the global epidemiology of hantavirus—particularly the Andes virus—have elevated concern among public health organizations and research laboratories worldwide.
Once considered a regional zoonotic risk, Andes virus is now drawing international attention due to documented human-to-human transmission and a series of travel-linked cases, including a notable 2026 cruise-associated cluster.
Hantaviruses are rodent-borne pathogens that can cause severe disease in humans. In the Americas, infection may lead to hantavirus cardiopulmonary syndrome (HCPS), which carries a reported case fatality rate of up to 38%.
Early symptoms are often nonspecific and may include:
  • Fever
  • Fatigue
  • Muscle aches
As disease progresses, patients may develop:
  • Gastrointestinal symptoms
  • Respiratory distress
  • Rapid clinical deterioration
Because symptoms overlap with influenza and other respiratory infections, molecular testing is critical for accurate detection and monitoring.
The Need for Rapid, Reliable Detection
Global health authorities, including the WHO, emphasize RT-PCR as a key method for hantavirus laboratory confirmation, alongside serology and immunohistochemistry.
The increase in travel-linked cases highlights the growing need for:
  • Rapid molecular detection
  • Scalable workflows
  • Broad laboratory compatibility
  • Reliable assay performance
Our Rapid Response Solution
In response to the 2026 incident, a molecular research solution was rapidly developed to support laboratories investigating hantavirus. Within days of the latest Andes virus sequence release, a new assay (AP7D4WY) was designed and optimized to align fully with the current incident strain.
Two RUO assay options are now available to support continuity and flexibility:
  • AP7D4WY: A newly designed, sequence-optimized assay aligned with the latest 2026 Andes virus strain and recommended for current incident detection
  • APU7ANW: An established, reference-based assay offering broad strain coverage and maintained sensitivity
This dual-assay approach enables laboratories to balance immediate incident responsiveness with broader surveillance needs. Early evaluation of this new assay is underway in U.S. public health laboratories, with results expected shortly.
Assay Comparison
FeatureAP7D4WY (new design)APU7ANW (existing design)
Design TypeSequence-optimized designReference-based design
CoverageAligned to current 2026 strainBroad coverage across known strains
PositioningRecommended for current incident detectionSuitable for broader surveillance
PerformanceOptimized for latest 2026 Andes virus sequenceMaintains sensitivity despite minor mismatch
Best UseEmerging incident responseEstablished strain monitoring
AP7D4WY is the recommended option for current incident detection because it is aligned to the latest publicly available Andes virus sequence.
APU7ANW remains a valuable option for broader hantavirus surveillance and continuity testing workflows.
A Scalable RT-qPCR Workflow
The workflow is designed for ease of implementation and scalability across multiple laboratory environments.
Our solution is built around a streamlined, end-to-end qPCR workflow designed for ease of adoption and scalability:
  1. Sample receipt and preparation
  2. RNA extraction using kits such as the MagMAX™ Prime Viral/Pathogen system 
  3. Real-time RT-qPCR using optimized master mixes such as TaqPath™ 1-Step RT-qPCR Master Mix, CG or TaqPath™ DuraPlex™ 1-Step RT-qPCR Master Mix as an option for multiplex workflows
  4. Custom TaqMan Gene Expression assays  including newly developed hantavirus-specific designs (e.g. AP7D4WY) that target the Andes orthohantavirus M segment (glycoprotein gene)
  5. Data analysis for qualitative detection or viral load quantification
The workflow incorporates internal and positive controls to help ensure reliability and reproducibility, and it is designed for ease of implementation and scalability across multiple laboratory environments. It is compatible with a wide range of real-time PCR instruments, enabling laboratories to integrate seamlessly with existing infrastructure and minimal disruption.
Supporting Laboratory Readiness
To support rapid deployment, key reagents—including assays, extraction kits, controls, and master mixes—have been proactively manufactured and made available for ordering.
Automated and manual sample preparation options are available to support:
  • High-throughput testing laboratories
  • Budget-conscious laboratories
  • Research-focused workflows
  • Flexible operational requirements
In addition, Thermo Fisher Scientific has collaborated with APHL to provide research kits to members to facilitate rapid workflow setup.
Enabling a Coordinated Global Response
The speed of this response reflects collaboration across R&D, assay design, operations, regulatory, supply chain, and commercial teams.
From sequence release to assay availability, the timeline demonstrates the importance of rapid scientific collaboration in addressing emerging infectious disease threats.
As the situation evolves, continued research and surveillance will remain essential to improving hantavirus detection strategies and incident preparedness.
Frequently Asked Questions
AP7D4WY is the recommended assay for the current 2026 Andes virus incident because it is aligned with the latest publicly available incident strain sequence.
AP7D4WY is optimized specifically for the current incident strain, while APU7ANW is a broader reference-based assay designed for wider strain coverage and continuity testing.
The workflow is well suited for public health laboratories, surveillance programs, academic research labs, and high-throughput molecular testing facilities with a related research focus.
Yes. The workflow is compatible with multiple commonly used RT-qPCR systems, including QuantStudio™, StepOne™, ViiA™, and Applied Biosystems™ 7500 platforms.
Yes. Automated extraction platforms such as the KingFisher™ Apex and Flex systems support scalable sample preparation for higher-throughput workflows.
Reference
CDC Hantavirus Information:
https://www.cdc.gov/hantavirus/about/index.html
For Research Use Only (RUO). Not for use in diagnostic procedures.
© 2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified. TaqMan is a trademark of Roche Molecular Systems, Inc., used under permission and license.
Geoffrey Cassell PHD, MBA
Written by:
Geoffrey Cassell, PhD, MBA
Senior Global Market Development Manager, GSD, Thermo Fisher Scientific
Geoffrey Cassell is a scientist-turned-marketing professional with extensive experience translating complex molecular biology technologies into impactful global marketing strategies. He earned his PhD in Cell and Molecular Biology from the University of California San Diego and San Diego State University, and later completed an MBA, building a foundation that bridges deep scientific expertise with business and commercial acumen. He has authored 9 peer-reviewed journal articles in publications such as Journal of Virology, Journal of Molecular Biology, and Nucleic Acids Research.
Read more Cassell, Geoffrey
Rob Eardley
Written by:
Senior Product Manager, GSD, Thermo Fisher Scientific
Rob Eardley is a Senior Product Manager at Thermo Fisher Scientific, specializing in qPCR assays and molecular research solutions. He leads cross-functional efforts to develop rapid response workflows for emerging infectious diseases, working closely with R&D, bioinformatics, and public health laboratories to support timely and scalable pathogen detection.
Read more Eardley, Rob