MagMAX™ Prime Viral/Pathogen NA Isolation Kit
Applied Biosystems™

MagMAX™ Prime Viral/Pathogen NA Isolation Kit

The MagMAX Prime Viral/Pathogen NA Isolation Kit is a magnetic-bead-based nucleic acid purification kit for the isolation and purification of viral, bacterial, yeast, and fungal nucleic acid from biological specimens.
Have Questions?
Catalog NumberQuantity
A58145Up to 600 Reactions
Catalog number A58145
Price (USD)
1,203.65
Online exclusive
1,254.00
Save 50.35
Each
Quantity:
Up to 600 Reactions
Price (USD)
1,203.65
Online exclusive
1,254.00
Save 50.35
Each

The MagMAX Prime Viral/Pathogen NA Isolation Kit is a magnetic-bead-based nucleic acid purification kit for the isolation and purification of viral, bacterial, parasitic, and fungal nucleic acid from human biological specimens.

Features of the MagMAX Prime Viral/Pathogen NA Isolation Kit include:

  • Designed to isolate and purify microbial nucleic acid from a range of biological samples such as swabs in transport media, saliva, stool, urine, plasma, and serum
  • Flexible workflow allowing various sample types to be batched on a single plate
  • Streamlined automation-ready protocols designed for basic and advanced lysis and advanced stool workflows
  • Fast automated KingFisher Flex, KingFisher Apex, KingFisher Apex Dx (RUO mode) scripts to allow for 96 samples to be processed in less than 60 minutes
  • Flexible protocol accommodates sample volume inputs of 200–400 μL
  • Elution volume of 200 μL for stool samples and 60 μL for all other sample types
  • Available lysis buffers and bead-beating accessories to extract nucleic acids from challenging organisms and raw/stabilized stool sample type

The kit is intended for use by qualified and trained clinical laboratory personnel specifically instructed and trained in automated magnetic bead purification techniques.

Compatible with Thermo Fisher infectious disease research assays

Have confidence that your nucleic acid extraction kit will work with your downstream application and enjoy a complete solution from Thermo Fisher Scientific.

Benefits of MagMAX magnetic bead–based purification

Magnetic beads offer many benefits compared to other technologies for isolating nucleic acid. Beads bind the nucleic acid more efficiently than glass-fiber filters, resulting in higher and more consistent yields. Additionally, because filters and vacuum manifolds are not used, there is no risk of filter clogging due to cellular particulates in samples. This clogging issue is of particular concern with protein-rich, large-volume samples such as whole blood or plasma that are commonly used for viral testing.

Accessories available for advanced workflow

Advanced lysis workflow

MagMAX Prime G+ Bacterial and Fungal Lysis Buffer (Cat. No. A59053)

Advanced stool workflow

MagMAX Prime Stool Lysis Buffer (Cat. No. A58154)

MagMAX Prime Bead Beating Tubes (Cat. No. A58155)

Standalone reagents

MagMAX Prime Viral/Pathogen Binding Beads (Cat. No. A58147)

MagMAX Prime Viral/Pathogen Elution Buffer (Cat. No. A58148)

MagMAX Prime Viral/Pathogen Wash I Solution (Cat. No. A58149)

MagMAX Prime Viral/Pathogen Binding Solution (Cat. No. A58150)

MagMAX Prime Viral/Pathogen Proteinase K (Cat. No. A58151)

MagMAX Prime Viral/Pathogen Proteinase K Dye (Cat. No. A58152)

For Research Use Only.
Specifications
For Use With (Application)Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
High-throughput CompatibilityHigh-throughput Compatible, Automated Protocols
Key FunctionsIsolate and purify viral, bacterial, parasitic, and fungal nucleic acid
No. of ReactionsUp to 600 Reactions
Nucleic Acid TypeDNA/RNA
Product TypeReagent
QuantityUp to 600 Reactions
Sample TypeSwabs in Transport Media, Saliva, Urine, Serum, Plasma
Sample Volume200–400 μL
Shipping ConditionRoom Temperature
TargetViral, Bacterial, Parasitic, and Fungal Nucleic Acid
FormatKit
Isolation TechnologyMagnetic Bead
Unit SizeEach
Contents & Storage

• 160 mL Binding Solution
• 300 mL Wash I Solution
• 6 mL Binding Beads
• 3 mL Proteinase K
• 36 mL Elution Buffer
• 30 μL Proteinase K dye

Store at 15–25°C.

Citations & References (7)

Citations & References
Abstract
Microbiota-derived short-chain fatty acids mediate Candida albicans gastrointestinal colonization resistance.
Authors:Mishra AA,Palmer SN,Zarek CM,Coughlin LA,Poulides N,Dakour J,Chiu RS,Sabaeifard P,Li W,Morrill AC,Kim J,Gan S,Behrendt CL,Antczak MI,Zhan X,Winter SE,Ready JM,Hooper LV,Koh AY
Journal:Cell host & microbe
PubMed ID:42242208
The gut microbiota plays critical roles in constraining Candida albicans colonization of the gastrointestinal (GI) tract, which is a key precursor to disseminated fungal infection in immunocompromised hosts. Depletion of commensal microbiota, such as by antibiotic treatment, increases C. albicans burden and promotes dissemination, yet the mechanisms of colonization resistance ... More
Streamlined CRISPR-based assays for detection and subtyping of avian influenza.
Authors:Huang Y,Guo A,Adams G,Lemieux JE,Myhrvold C
Journal:medRxiv : the preprint server for health sciences
PubMed ID:40909817
Avian influenza viruses (AIVs) are zoonotic pathogens that pose an increasing global threat due to their potential for significant economic losses in agriculture, spillover into humans, and the risk of a pandemic should human-to-human transmission occur. These concerns underscore the need for rapid, sensitive and specific tools to detect and ... More
Establishing Methods to Monitor Influenza (A)H5N1 Virus in Dairy Cattle Milk, Massachusetts, USA.
Authors:Stachler E,Gnirke A,McMahon K,Gomez M,Stenson L,Guevara-Reyes C,Knoll H,Hill T,Hill S,Messer KS,Arizti-Sanz J,Albeez F,Curtis E,Samani P,Wewior N,O'Connor DH,Vuyk W,Khoury SE,Schnizlein MK,Rockey NC,Broemmel Z,Mina M,Madoff LC,Wohl S,O'Connor L,Brown CM,Ozonoff A,Park DJ,MacInnis BL,Sabeti PC
Journal:Emerging infectious diseases
PubMed ID:40138725
Highly pathogenic avian influenza A(H5N1) virus has caused a multistate outbreak among US dairy cattle, spreading across 16 states and infecting hundreds of herds since its onset. We rapidly developed and optimized PCR-based detection assays and sequencing protocols to support H5N1 molecular surveillance. Using 214 retail milk samples from 20 ... More
Evaluation of Self-Collected Mouth Rinse Specimens for Coronavirus Disease 2019 Testing: A Pilot Study
Authors:Kento Fukano; Junko S. Takeuchi; Azusa Kamikawa; Wataru Sugiura
Journal:COVID
PubMed ID:
Saliva specimens are widely used for coronavirus disease 2019 (COVID-19) testing using RT-qPCR due to their advantages over nasopharyngeal swabs of being non-invasive and self-collectable. However, saliva collection can be time-consuming in individuals with reduced saliva secretion, including those with diabetes, diseases involving salivary glands such as Sjögren’s syndrome, and ... More
Interdependent RNA structural motifs at the 3'-terminus of the West Nile virus genome regulate viral growth.
Authors:Tsao LH,Brackney DE,Pyle AM
Journal:bioRxiv : the preprint server for biology
PubMed ID:41959387
The RNA genome of West Nile Virus (WNV) folds into an elaborate series of RNA structural elements that are crucial for viral function. Among these elements, four pseudoknots (PKs) at the viral 3’-terminus, designated as SLII, SLIV, DBI, and DBII, are among the most crucial players in the overall flaviviral ... More